Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Product Lines
New PCR Products
Key Resources
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 Probe based Quantitative PCR
PCR
Probe based QPCR relies on the sequence–specific detection of a desired PCR product. Unlike SYBR based QPCR methods that detect all double–stranded DNA, probe based QPCR utilizes a fluorescent–labeled target-specific probe resulting in increased specificity and sensitivity. Additionally, a variety of fluorescent dyes are available so that multiple primers can be used to simultaneously amplify many sequences.
  • Ideal for high throughput. ReadyMixes contain all necessary components for QPCR you simply add the fluorescent detection chemistry, primers, and template.
  • Specific – JumpStart Taq antibody prevents non–specific product formation through it hot–start mechanism.
  • Maximum flexibility in detection method, since no detection method has been incorporated into formulation.
  • Optimized formulations with the addition of dUTP to facilitate carry–over prevention from previous PCR.
Product Name Technical Bulletin Product # 
JumpStart Taq ReadyMix for for High Throughput QPCR D6442
JumpStart Taq ReadyMix for QPCR D7440
JumpStart Taq ReadyMix with dUTP D9191
Mechanism of probe based protocol
Click Image to View Figure in its Entirety Figures 1 & 2. Superior Amplification Efficiency with Lower CT.
Sigma-Genosys, Sigma-Proligo
Probe–based real–time PCR, dual labeled probes and Molecular Beacons are available from Sigma Genosys.
Sigma-Aldrich Teams with Epoch Biosciences
Sigma-Aldrich has teamed with Epoch Biosciences to create an optimized PCR reagent for the MGB Eclipse™ Probe Systems for both Gene Expression Profiling and SNP Analysis. The PCR reagents are pre-formulated with necessary buffers and deoxynucleotides and when combined with JumpStart™ Taq DNA Polymerase creates a ready-to-use 2X PCR master mix.
Back to Quantitative PCR home page.
Routine Amplification | Human Genomic DNA | Long & Accurate PCR Reagents | Hot Start PCR | RT-PCR | Quantitative PCR | Specialty Enzymes | Ultrapure Nucleotides | PCR Reaction Components | PCR Lab Equipment | DNA/RNA Molecular Weight Markers | Whole Genome Amplification