Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Product Lines
New PCR Products
Key Resources
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 SYBR® Green based Quantitative PCR
PCR
SYBR Green I, a commonly used fluorescent DNA binding dye, binds all double–stranded DNA and detection is monitored by measuring the increase in fluorescence throughout the cycle. SYBR Green I has an excitation and emission maxima of 494 nm and 521 nm, respectively. Specificity of Sigma's SYBR based QPCR detection is greatly enhanced by the incorporation of a hot–start mediated taq polymerase, JumpStart Taq.
  • Convenient – Delivers the benefits of antibody–inactivated hot–start PCR with SYBR Green detection in a ReadyMix ideal for high throughput applications; only primers and template are required.
  • Specific – JumpStart Taq antibody prevents non–specific product formation through its hot–start mechanism.
  • Flexible – SYBR Green JumpStart Taq ReadyMixes for SYBR based QPCR are formulated with MgCl2 or packaged with a separate vial for ease of optimization. Additionally, our ReadyMixes are compatible with tube– and plate–based instruments.
Product Name Technical Bulletin Product # 
SYBR Green JumpStart Taq ReadyMix for High Throughput QPCR S9194
SYBR Green JumpStart Taq ReadyMix for QPCR S4438
SYBR Green JumpStart Taq ReadyMix without MgCl2 S5193
Mechanism of SYBR Green–based detection
Click Image to View Figure in its Entirety Figure 1. Linearity of pBAC–2cp 10–fold dilutions with SYBR Green JumpStart Taq ReadyMix.
Click Image to View Figure in its Entirety Figure 2. CT Values for Lambda Amplicon using SYBR Green I JumpStart Taq ReadyMix.
Back to Quantitative PCR home page.
Routine Amplification | Human Genomic DNA | Long & Accurate PCR Reagents | Hot Start PCR | RT-PCR | Quantitative PCR | Specialty Enzymes | Ultrapure Nucleotides | PCR Reaction Components | PCR Lab Equipment | DNA/RNA Molecular Weight Markers | Whole Genome Amplification