Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Product Lines
New PCR Products
Key Resources
Whole Genome Amplification
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 PCR Specialty Enzymes:
 Polymerases for DNA Repair and improved amplification
PCR
We offer specialty DNA polymerases optimized to increase amplification efficiency of targeted bacterial sequences and to repair and restore damaged or degraded DNA. Our most up-to-date products are MTP Taq DNA Polymerase and Restorase DNA Polymerase.
Sigma's innovative specialty enzymes can be used to amplify bacterial DNA or repair damaged DNA. The MTP™ Taq DNA Polymerase is ideal for researchers detecting and identifying bacterial DNA, looking for a more accurate method in mutation scanning techniques, or wanting to prevent the amplification of undesired DNA sequences. This product is purified through a proprietary process to minimize the levels of contaminating DNA, which in turn, accurately amplifies targeted conserved sequences such as the 16S rRNA region. Our MTP Taq undergoes strict quality control testing to ensure the absence of detectable levels of contaminating DNA.
The Restorase™ DNA Polymerase contains a DNA repair enzyme that facilitates repair and amplification of damaged DNA. Many times, DNA templates are compromised when damaged by exposure to acid, alkylating agents, heat or light. These damages block the progression of DNA polymerase, thereby affecting PCR efficiency. Restorase functions by modifying the damaged sites allowing subsequent template copying. The Restorase DNA Polymerase is ideal for researchers who are unable to achieve amplification of damaged DNA templates when using other thermostable DNA polymerases. This enables researchers to work with damaged templates that would otherwise be abandoned.
Products:
MTP™ Taq DNA Polymerase (D7442)
Restorase™ DNA Polymerase (R1028)
back to top
back to PCR home page
Routine Amplification | Human Genomic DNA | Long & Accurate PCR Reagents | Hot Start PCR | RT-PCR | Quantitative PCR | Specialty Enzymes | Ultrapure Nucleotides | PCR Reaction Components | PCR Lab Equipment | DNA/RNA Molecular Weight Markers | Whole Genome Amplification