|
SigmaAldrich's Ultrapure Nucleotides are available as individual nucleotides or mixes. Each product is tested for suitability in PCR and DNase and RNase free.
- Ultrapure dNTPs – Minimum > 99% determined by HPLC
- Minimize Contamination Risk – Certified DNase & RNase free
- Convenient Premixed dNTP Solution Equimolar amounts of each dNTP means less pipetting
| 10mM
solutions | 100mM solutions |
| dNTP | Product Number | Pack Size | Product Number | Pack Size |
| dATP |
D6920 | 0.5 ml (5 µmol) |
D4788 | 0.25 ml (25µmol) |
| dCTP |
D7045 | 0.5 ml (5 µmol) |
D4913 | 0.25 ml (25µmol) |
| dGTP |
D7170 | 0.5ml (5 µmol) |
D5038 | 0.25ml (25µmol) |
| TTP |
T7791 | 0.5ml (5 µmol) |
T9656 | 0.25ml (25µmol) |
dNTP Set 1 vial of each
dNTP |
DNTP-10 | 0.5 ml (5 µmol) of each
dNTP |
DNTP-100
DNTP-100A | 0.25 ml (25 µmol) of each
dNTP 1.00 ml (100 µmol) of each dNTP
|
dNTP Mix Solution of all four
dNTPs |
D7295 | 0.2 ml (2 µmol)
0.5 ml (5 µmol) 20 x 0.2ml (20x2 5mol) |
Routine Amplification |
Human Genomic DNA |
Long & Accurate PCR Reagents |
Hot Start PCR |
RT-PCR |
Quantitative PCR |
Specialty Enzymes |
Ultrapure Nucleotides |
PCR Reaction Components |
PCR Lab Equipment |
DNA/RNA Molecular Weight Markers |
Whole Genome Amplification
|