Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
GenomePlex Products
Key Resources
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 GenomePlex® Technology Overview
Whole Genome Amplification
Move Beyond Traditional PCR Limitation...Unsurpassed Yield,
Unlimited Potential
The GenomePlex® Whole Genome Amplification (WGA) family of products provides a robust and accurate method of amplifying nanogram quantities of starting material into microgram yields with minimal allele drop out.
The GenomePlex WGA product line utilizes a proprietary amplification technology based upon random fragmentation of genomic DNA and conversion of the resulting small fragments to PCR-amplifiable OmniPlex® Library molecules flanked by universal priming sites. The OmniPlex library is then PCR amplified using universal oligonucleotide primers and a limited number of cycles.
WGA has been used in a variety of applications,1, 2, 3 and is suitable for use with purified genomic DNA from a variety of sources including blood cards, whole blood, buccal swabs, soil, plant, and formalin-fixed paraffin-embedded (FFPE) tissues. GenomePlex WGA uses nanogram quantities of starting genomic DNA, which after PCR yields 5 to 10 µg of WGA product. After purification, the WGA product can be analyzed in a manner similar to any genomic or chromosomal DNA sample. A number of downstream applications may be performed including TaqMan® assays, CGH analysis, SNP analysis, sequencing, etc.
  • Choose from a variety of DNA sources: whole blood, blood card, plasma, serum, buccal swab, plant, soil, formalin-fixed paraffin-embedded (FFPE) tissue and single cells
  • See a complete representation of the entire genome with no detectable allele bias.
  • Increase amplification accuracy with WGA DNA Polymerase
  • Preserve precious source material by amplifying nanogram amounts of starting genomic DNA into microgram yields in a few hours
  • Enjoy the compatibility GenomePlex provides with many downstream applications: TaqMan assays, SNP analysis, microarray analysis, sequencing, or store at -20 °C for future use
References
  1. Little, S.E., et al., Array CGH using whole genome amplification of fresh-frozen and formalin-fixed, paraffin-embedded tumor DNA. Genomics. Oct 31, 2005.
  2. Barker, D. L., et al., Two methods of whole-genome amplification enable accurate genotyping across a 2320-SNP linkage panel. Genome Res., 14, 901-907 (2004).
  3. Gribble, S., et al., Chromosome paints from single copies of chromosomes. Chromosome Res., 12, 143-151 (2004).
  4. Thorstenson, Y. R., et al., An Automated Hydrodynamic Process for Controlled, Unbiased DNA Shearing. Genome Res., 8, 848-855 (1998).
  5. Park, J.W., et al., Comparing Whole-Genome Amplification Methods and Sources of Biological Samples for Single-Nucleotide Polymorphism Genotyping. Clinical Chemistry., 51(8), 1520-1523 (2004).
back to the Whole Genome Amplification home page