Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
GenomePlex Products
Key Resources
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 GenomePlex® Single Cell WGA Genotyping Data
PCR
Accurate Amplification Preserving Genome Representation
Single Cell WGA (WGA4) Preserves Genotype of the Original Genome.
Single Cell Whole Genome Amplification Genotyping Data Female sample
Female sample
Positive control
Negative control
Single Cell WGA was performed on amniotic fluid sourced fibroblast cells drawn from two different patients. The genotyping control samples provide no signals in the negative control and male specific peaks for the positive control. The two upper graphs represent data for two different female samples as amplified with the single cell WGA kit. These data indicate that GenomePlex single cell amplified products are compatible with gender study assays.
back to Single Cell Whole Genome Amplification Kit