Molecular Biology
Automated Sequencing
Cloning and Expression
DNA and RNA Purification
Gene Expression Analysis
Molecular Biology Reagents
Nucleic Acid Detection
Nucleic Acid Electrophoresis
PCR
Whole Genome Amplification
GenomePlex Products
Key Resources
New Products
Neuroscience
Nutrition Research
Obesity Research
Peptides and Proteins
Plant Biotechnology
Proteomics and Protein Expr.
Stable Isotopes
Stem Cell Biology
Your Favorite Gene - Search
Life Science Innovations
PathFinder
 GenomePlex® Single Cell WGA Kit STR Application Data
PCR
Single Cell WGA (WGA4) Supports Robust Amplification
Single Cell Whole Genome Amplification STR Data
Lane 1: Marker
Lane 2: Positive Control
Lane 3: Negative Control
Lane 4: cell 1
Lane 5: cell 2
Lane 6: cell 3
Lane 7: cell 4
Lane 8: cell 5
Five fresh lymphocytes were lysed and amplified using the Single Cell WGA Kit (WGA4). The measure of success was performed with the DXS1623 microsatellite marker, amplified by two-rounds PCR. The final product was verified on a 1.5% agarose gel. No contamination was noted in the negative control sample.
Data is courtesy of Dr. Christophe Beroud from Laboratoire de Genetique Moleculaire et Chromosomique, Institute Universitaire de Recherche Clinique, France.
back to Single Cell Whole Genome Amplification Kit