Ethyl Vinyl Acetate Bags
Polyethylene Bags
Rigging Sets
Components
Tank Liners
Tubing
Storage and Transport
General Information
Ethyl Vinyl Acetate Bags
On this Page
BIOEAZE Bags - Ethyl Vinyl Acetate < 5 L
BIOEAZE Bags - Ethyl Vinyl Acetate 5 - 10 L
BIOEAZE Bags - Ethyl Vinyl Acetate 10 - 20 L
BIOEAZE Bags - Ethyl Vinyl Acetate < 5 L
Catalog #
Name
1522B
BIOEAZE 350 mL Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 1 × 1 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 1 Medicinal Port
· 1 Spike Port
Individually polybagged
>
BIOEAZE 350 mL EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1935B
BIOEAZE 350 mL Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed with:
· 2 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 2 × 1/4" Male Quick Connect
· 1 Spike Port
Individually polybagged
>
BIOEAZE 350 mL EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1446B
BIOEAZE 350 mL Ethyl Vinyl Acetate (EVA) Sterile Bag 10/pk
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 1 × 1.5 ft. length of 1/8" × 1/4" Clear 082 C-Flex® Tubing
· 1 Spike Port
· 1 Swabable Needleless Luer Valve
Individually polybagged
>
BIOEAZE 350 mL EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1521B
BIOEAZE 1 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed for Terumo® Welding to 1331B, 1332B and 1333B
BIOEAZE
Polyethylene (PE) Sterile Bags
Designed with:
· 1 × 1 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 1 Medicinal Port
· 1 Spike Port
Individually polybagged
>
BIOEAZE 1 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1936B
BIOEAZE 1 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed with:
· 2 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 2 × 1/4" Male Quick Connect
· 1 Spike Port
Individually polybagged
>
BIOEAZE 1 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1937B
BIOEAZE 2 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed with:
· 2 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 2 × 1/4" Male Quick Connect
· 1 Spike Port
Individually polybagged
>
BIOEAZE 2 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1933B
BIOEAZE 2 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 1 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 1/4" Male Quick Connect
· 1 × 3 ft. length of 1/8" × 1/4" Clear C-Flex® Tubing
· 1 Male Luer Lock with Female Cap
· 1 Spike Port
Individually polybagged
>
BIOEAZE 2 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
For ordering information please contact
your local SAFC Biosciences office
back to top
BIOEAZE Bags - Ethyl Vinyl Acetate 5 - 10 L
Catalog #
Name
1934B
BIOEAZE 5 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed for use with a Terumo® Aseptic Connecting Device
Designed with:
· 1 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 1/4" Male Quick Connect
· 1 × 3 ft. length of 1/8" × 1/4" Clear C-Flex® Tubing
· 1 Male Luer Lock with Female Cap
· 1 Spike Port
Individually polybagged
>
BIOEAZE 5 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1938B
BIOEAZE 5 L Ethyl Vinyl Acetate (EVA) Sterile Bag 5/pk
Designed with:
· 2 × 6" length of 1/4" × 3/8" Clear C-Flex® Tubing
· 2 × 1/4" Male Quick Connect
· 1 Spike Port
Individually polybagged
>
BIOEAZE 5 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1523B
BIOEAZE 5 - 10 L Ethyl Vinyl Acetate (EVA) Sterile Bag
Designed with:
· 2 × 2.5 ft. length of Size 15 Opaque 001 C-Flex® Tubing
· 1/4" Male and Female Quick Connects with Caps
· 1 Spike Port
Double polybagged
>
BIOEAZE 5 - 10 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1524B
BIOEAZE 5 - 10 L Ethyl Vinyl Acetate (EVA) Sterile Bag
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 2 × 2.5 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 1 Spike Port
Double polybagged
>
BIOEAZE 5 - 10 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
For ordering information please contact
your local SAFC Biosciences office
back to top
BIOEAZE Bags - Ethyl Vinyl Acetate 10 - 20 L
Catalog #
Name
1329B
BIOEAZE 10 - 20 L Ethyl Vinyl Acetate (EVA) Sterile Bag
Designed for use with a Terumo® Aseptic Connecting Device (ACD)
Designed with:
· 2 × 2.5 ft. length of Size 16 Opaque 001 C-Flex® Tubing
· 1 Spike Port
Double polybagged
>
BIOEAZE 10 - 20 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
1330B
BIOEAZE 10 - 20 L Ethyl Vinyl Acetate (EVA) Sterile Bag
Designed with:
· 2 × 2.5 ft. length of Size 15 Opaque 001 C-Flex® Tubing
· 1/4" Male and Female Quick Connects with Caps
· 1 Spike Port
Double polybagged
>
BIOEAZE 10 - 20 L EVA Bag
>
BIOEAZE Bags EVA
>
Chemical Compatibility For Bag Films
>
TOC Testing of CC Water Stored in BIOEAZE Bags
For ordering information please contact
your local SAFC Biosciences office
back to top