|
Rigging Sets |
|
| |
|
| Catalog # |
Name |
| 1336R |
BIOEAZE Rigging Set 1/4" with Male-Female Quick Connects Sterile
C-Flex® Tubing, Size 15 Opaque 001
· with a 1/4" Male Quick Connect
· with a Cap at one end and a Female Quick Connect at the other end
Individually double polybagged
10 ft. length
> BIOEAZE Rigging Set
|
| 1337R |
BIOEAZE Rigging Set 3/8" with Male-Female Quick Connects Sterile
C-Flex® Tubing, Size 73 Opaque 001
· with a 3/8" Male Quick Connect
· with a Cap at one end and a Female Quick Connect at the other end
Individually double polybagged
10 ft. length
> BIOEAZE Rigging Set
|
| 1520R |
BIOEAZE Rigging Set 3/8" with Male-Female Quick Connects Sterile
C-Flex® Tubing, Size 73 Opaque 001
· with a 3/8" Male Quick Connect
· with a Cap at one end and a Female Quick Connect at the other end
Individually double polybagged
25 ft. length
> BIOEAZE Rigging Set
|
| Catalog # |
Name |
| 1339R |
BIOEAZE Rigging Set Aseptic Connecting Device (ACD) Clear Sterile
C-Flex® Tubing, 1/8" x 1/4" Clear 082-C
· for use with Aseptic Connecting Device (ACD)
· with plugs at each end
Individually double polybagged
10 ft. length
> BIOEAZE Rigging Set
|
| 1338R |
BIOEAZE Rigging Set Aseptic Connecting Device (ACD) Sterile
C-Flex® Tubing, Size 16 Opaque 001
· for use with Aseptic Connecting Device (ACD)
· with plugs at each end
Individually double polybagged
10 ft. length
> BIOEAZE Rigging Set
|
| Catalog # |
Name |
| 1654R |
BIOEAZE Dispensing Tube Set Sterile
· with Male Quick Connect
Individually double polybagged
2 ft. length
> BIOEAZE Tube Set
|
| 1655R |
BIOEAZE Dispensing Tube Set Sterile
· with Aseptic Connecting Device (ACD) Weldable Size 16 Opaque 001 C-Flex® Tubing
Individually double polybagged
5 ft. length
> BIOEAZE Tube Set
|
| 1656R |
BIOEAZE Dispensing Tube Set Sterile
· with Aseptic Connecting Device (ACD) Weldable 1/8" x 1/4" Clear 082 C-Flex® Tubing
Individually double polybagged
5 ft. length
> BIOEAZE Tube Set