|
Insect Cell Culture Media |
|
| |
|
| Catalog # |
Name |
| 51942C |
Hink's TNM-FH Insect Medium
supplemented Grace's, with 4.1 mM L-glutamine, with 3.33 g/L lactalbumin hydrolysate (LAH), with 3.33 g/L ultrafiltered yeastolate
Liquid Medium (LM)
> Hink's TNM-FH Insect Medium
> MSDS - US Version
> MSDS - European Version
Available Sizes: 500 mL, 1000 mL
Storage Conditions: 2 to 8 °C