|
Detachment Factors |
|
| |
|
| Catalog # |
Name |
| 59427C |
Trypsin Solution 10X
2.5% trypsin, trypsin gamma irradiated by SER-TAIN Process, in Hanks' Balanced Salt Solution
Frozen Solution
> Trypsin 10X
> MSDS - US Version
> MSDS - European Version
Available Sizes: 100 mL, 500 mL
Storage Conditions: -10 to -40 °C
|
| Catalog # |
Name |
| 59428C |
Trypsin-EDTA Solution 1X
0.25% trypsin, 0.02% EDTA, trypsin gamma irradiated by SER-TAIN Process, in Hanks' Balanced Salt Solution
Frozen Solution
> Trypsin-EDTA 1X
> MSDS - US Version
> MSDS - European Version
Available Sizes: 100 mL, 500 mL
Storage Conditions: -10 to -40 °C
|
| 59429C |
Trypsin-EDTA Solution 1X
0.25% trypsin, 0.1% EDTA, trypsin gamma irradiated by SER-TAIN Process, in Hanks' Balanced Salt Solution
Frozen Solution
> Trypsin-EDTA 1X
> MSDS - US Version
Available Sizes: 100 mL, 500 mL
Storage Conditions: -10 to -40 °C
|
| 59430C |
Trypsin-EDTA Solution 1X
0.12% trypsin, 0.02% EDTA,trypsin gamma irradiated by SER-TAIN Process, without phenol red, in Dulbecco's Phosphate Buffered Saline
Frozen Solution
> MSDS - US Version
> MSDS - European Version
Available Sizes: 100 mL, 500 mL
Storage Conditions: -10 to -40 °C
|
| 59417C |
Trypsin-EDTA Solution 1X
0.05% trypsin, 0.02% EDTA, trypsin gamma irradiated by SER-TAIN Process, in Hanks' Balanced Salt Solution
Frozen Solution
> Trypsin-EDTA 1X
> MSDS - US Version
Available Sizes: 100 mL, 500 mL
Storage Conditions: -10 to -40 °C
|
| Catalog # |
Name |
| 59418C |
Trypsin-EDTA Solution 10X
0.5% trypsin, 0.2% EDTA, trypsin gamma irradiated by SER-TAIN Process, without phenol red, in saline
Frozen Solution
> Trypsin - EDTA (10X Solution) (0.5% Trypsin, 0.2% EDTA)
Available Sizes: 100 mL
Storage Conditions: -10 to -40 °C
|
| Catalog # |
Name |
| 59226C |
NO-ZYME
PBS Base, without calcium, without magnesium, without phenol red
Liquid Solution
> NO-ZYME
> MSDS - US Version
Available Sizes: 100 mL
Storage Conditions: 15 to 30 °C