Growth Factors and Proteins Components
 On this Page
Albumin
Transferrin
Insulin
Growth Factors (EGF, IGF, TGF)
Albumin
Catalog # Name
85041C Bovine Serum Albumin - Fatty Acid Free
Lyophilized Powder
  > BSA - Fatty Acid Free
  > MSDS - US Version
  Available Sizes: 10 g, 50 g, 100 g, 500 g, 1 kg
  Storage Conditions: 2 to 8 °C
85040C Bovine Serum Albumin - Fraction V
Lyophilized Powder
  > BSA - Fraction V
  > MSDS - US Version
  Available Sizes: 10 g, 50 g, 100 g, 500 g, 1 kg
  Storage Conditions: 2 to 8 °C
  For ordering information please contact your local SAFC Biosciences office
back to top
Transferrin
Catalog # Name
85020C Bovine Transferrin - apo
essentially iron free
Lyophilized Powder
  > Bovine Transferrin - apo
  > MSDS - US Version
  Available Sizes: 100 mg, 1 g, 5 g, 25 g
  Storage Conditions: -10 to -40 °C
  For ordering information please contact your local SAFC Biosciences office
back to top
Insulin
Catalog # Name
91077C Insulin, Recombinant Human
chemically defined
Dry Powder
  > Insulin, Recombinant Human
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 100 mg, 250 mg, 1 g, 5 g, 10 g, 50 g
  Storage Conditions: -10 to -40 °C
  For ordering information please contact your local SAFC Biosciences office
back to top
Growth Factors (EGF, IGF, TGF)
Catalog # Name
85570C LONG®EGF Media Grade
Lyophilized Powder
  > LONG®EGF
  > MSDS - US Version
  Available Sizes: 1 mg, 5 mg, 10 mg, 20 mg, 50 mg
  Storage Conditions: 2 to 8 °C
85580C LONG®R³IGF-I Lyophilized, Media Grade
This product is intended for cell culture use only. When ordering this product you may be required to provide SAFC Biosciences evidence of the product's intended use.
Lyophilized Powder
  > LONG®R³IGF-I
  > LONG®R³IGF-I FAQs — Strategic Sourcing
  > LONG®R³IGF-I FAQs
  > The Science, Features and Benefits of LONG®R³IGF-I
  > LONG®R³IGF-I Storage, Stability and Specifications
  > MSDS - US Version
  Available Sizes: 1 mg, 5 mg, 10 mg, 20 mg, 50 mg
  Storage Conditions: 2 to 8 °C
91590C
LONG®R³IGF-I Liquid, Media Grade
This product is intended for cell culture use only. When ordering this product you may be required to provide SAFC Biosciences evidence of the product's intended use.
  1 mg/mL filtered solution in 100 mM acetic acid
  > LONG®R³IGF-I Liquid  
  > LONG®R³IGF-I FAQs — Strategic Sourcing
  > LONG®R³IGF-I FAQs
  > The Science, Features and Benefits of LONG®R³IGF-I
  > LONG®R³IGF-I Storage, Stability and Specifications
  > MSDS - US Version
  > MSDS - EU Version
  Available Sizes: 5 mL, 100 mL
  Storage Conditions: 2 to 8 °C
85550C LONG™TGF-α Media Grade
Lyophilized Powder
  > LONG®TGF-a
  > MSDS - US Version
  Available Sizes: 1 mg, 5 mg, 10 mg
  Storage Conditions: 2 to 8 °C
  For ordering information please contact your local SAFC Biosciences office
back to top