|
Reagents |
|
| |
|
| Catalog # |
Name |
| 59222C |
Sodium Chloride 5M
292.2 g/L sodium chloride
Liquid Solution
> Sodium Chloride 5M
> MSDS - US Version
Available Sizes: 500 mL, 1000 mL, 20 L
Storage Conditions: 15 to 30 °C |
| 59412C |
Hoechst Stain Solution
Hoechst stain solution, 1.2 g/L BisBenzimide H 33258
Liquid Solution
> Hoechst Stain
> MSDS - US Version
Available Sizes: 50 mL
Storage Conditions: -10 to -40 °C |
| 59415C |
Phenol Red Solution 0.5%
Phenol red solution, 5.0 g/L phenol red
Liquid Solution
> Phenol Red 0.5%
> MSDS - US Version
Available Sizes: 100 mL
Storage Conditions: 15 to 30 °C |
| 59410C |
Trypan Blue Solution 0.4%
Trypan blue solution, 4.0 g/L trypan blue, in saline
Liquid Solution
> Trypan Blue 0.4%
> MSDS - US Version
Available Sizes: 100 mL
Storage Conditions: 15 to 30 °C |
| 59900C |
Water - Cell Culture Grade
meets criteria for WFI, endotoxin ≤0.25 EU/mL
Liquid
> Water - Cell Culture Grade
> MSDS - US Version
Available Sizes: 500 mL, 1000 mL, 20 L
Storage Conditions: 15 to 30 °C |