Supplements
 On this Page
Supplements
Supplements
Catalog # Name
58980C Diamond Vitamin Tween™ 80 Solution 40X
Frozen Solution
  > Diamond Vitamin Tween™ 80 40X
  > MSDS - US Version
  Available Sizes: 100 mL, 500 mL
  Storage Conditions: -10 to -40 °C
90114C L-Glutamine Powder
Dry Powder
  > L-Glutamine Powder
  > MSDS - US Version
  Available Sizes: 25 g, 100 g, 500 g, 1 kg, 5 kg
  Storage Conditions: 15 to 30 °C
59202C L-Glutamine Solution 200 mM
L-glutamine solution 200 mM, 29.23 mg/mL in saline
Frozen Solution
  > L-Glutamine Solution
  > L-Glutamine Stability
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 100 mL, 500 mL
  Storage Conditions: -10 to -40 °C
59212C Glutamine-S 200 mM
Glutamine-S, L-alanyl-L-glutamine solution, 43.44 mg/mL in saline
Frozen Solution
  > Glutamine-S 200 mM
  > Glutamine-S Stability
  > MSDS - US Version
  Available Sizes: 100 mL, 500 mL
  Storage Conditions: -10 to -40 °C
58672C GS Supplement 50X
Frozen Solution
  > GS Supplement 50X
  > MSDS - US Version
  Available Sizes: 100 mL
  Storage Conditions: -10 to -40 °C
58901C Lactalbumin Hydrolysate Solution 50X
166.5 g/L LAH
Liquid Solution
  > Lactalbumin Hydrolysate 50X
  > MSDS - US Version
  Available Sizes: 100 mL
  Storage Conditions: 2 to 8 °C
14101C Lipid Concentrate 500X, Chemically Defined
Liquid Solution
  > Lipid Concentrate 500X, Chemically Defined
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 50 mL, 100 mL
  Storage Conditions: 2 to 8 °C
58252C MEM Amino Acids Solution 50X
Frozen Solution
  > MEM Amino Acids Solution 50X
  > MSDS - US Version
  Available Sizes: 100 mL
  Storage Conditions: -10 to -40 °C
58903C Soy Hydrolysate UF Solution 50X
250 g/L soy hydrolysate UF
Liquid Solution
  > Soy Hydrolysate UF 50X
  > MSDS - US Version
  > MSDS - European Version
  Available Sizes: 100 mL
  Storage Conditions: 2 to 8 °C
  For ordering information please contact your local SAFC Biosciences office
back to top