SAFC Hamburg, Germany
SAFC Supply Solutions
SAFC Hamsburg: a multi-purpose facility with built-in flexibility and the capability to manufacture a complex mix of products as well as prepare and purify alternative mixes of high volume standard products and small scale specialties.
DNA and RNA phosphoramidites are manufactured in glass-lined reactors and purified by preparative column chromatography on silica gel with medium-to-high pressure chromatography equipment.
What the SAFC Hamburg facility
can provide for you
Modern laboratories with state-of-the-art analytical support
Integrated solvent delivery
A strong background in specialty monomer manufacturing
CPG production using the Schott process
Extensive storage and warehousing facilities with worldwide distribution services
Contact SAFC Hamburg
SAFC Hamburg Facility in Germany
Quality Manager
Hamburg, Germany
Tel: +49-40-797-02-0
Fax: +49-40-797-02-100
Schedule a Site Audit
Request Quality Information
Certificates and Documents
ISO 9001:2000 Certification (124k PDF)
Certificates of Analysis
Product data RP HPLC, NMR
Tests performed on bulk product
UV/VIS base identity test performed on finished good product