|
SAFC Buchs is located in Buchs, Switzerland and encompasses 402,400 ft2 of manufacturing space. There are multiple production facilities within this ISO 9002 certified site that are responsible for the production of over 4,000 compounds.
The company recently committed to a $5.4 million cGMP production expansion to provide additional improvements in material and personnel flow and expand cGMP storage. The expansion is expected to be on line in the second quarter, 2008.
SAFC Buchs has a long history of producing high-quality precursors & actives. The plant specializes in complex, multi-step synthesis to 1,600 liters and low temperature reactions (-60°C) up to 400 liters. A broad range of production technologies and multi-purpose equipment offers batch sizes from grams to tons.
This facility operates under cGMP, certified by the Swiss authorities.
| |
Custom synthesis and process development |
| |
Production of high-quality intermediates and bulk actives |
| |
Technology strengths are aimed toward low-volume, highly complicated chemistries |
| |
Reactor capacity from 5 to 400 gallon (20 L - 1,600 L) glass-lined and Hastelloy® |
| |
Operating temperature range from -60°C up to +180°C |
| |
Safety of all processes is evaluated using state-of-the-art DSCRC 1 and ARC calorimeter equipment |
| |
|
| |
Industriestrasse 25
9470 Buchs
Switzerland
Swiss Free Call: 0800 80 00 80
Tel: +41 (0) 81 755 27 32
Fax: +41 (0) 81 755 27 70
E-Mail: hitechinfo@sial.com |
|
| |
|
| |
Bromination |
50500 GA |
| |
Chlorination |
50500 GA |
| |
Phosgenation |
50500 GA |
| |
Sulfonation |
50500 GA |
|
| |
ISO 9002 Certified |
| |
Biotransformations |
| |
Distillations |
| |
Complex multi-step synthesis |
|
| Equipment Summary |
| Vessels |
50200 GA
(200800 L) |
>200500 GA
(>8002,000 L) |
>5002,000 GA
(>2,0008,000 L) |
>2,000 GA
(>8,000 L) |
Total |
| Glass-Lined |
20 |
6 |
0 |
0 |
26 |
| Stainless Steel |
1 |
1 |
0 |
0 |
2 |
| Hastelloy |
0 |
0 |
0 |
0 |
0 |
| Temp. Range (°C) |
-60 to +225 |
-30 to +200 |
|
|
|
| Total Capacity |
1,330 GA
(5,320 L) |
2,165 GA
(8,660 L) |
|
|
3,495 GA
(13,980 L) |