EpiFill™
SAFC Hitech - Silicon Semiconductor
 EpiFill bulk delivery system
EpiFill Unit Door OpenThe ability to constantly replenish the chemicals in the EpiFill system results in decreased equipment downtime, minimized human traffic in clean-rooms, reduced safety hazards and higher overall throughput.
 Applications include
Liquid source delivery for production environment deposition systems employed to fabricate semiconductor devices
TMA/TMG delivery in MOVPE/MOCVD/ALD or solar cells, HBLEDSs
TMG delivery for MOVPE HBTs
TMA delivery for ALD high-k Al2O3
HfD-04/TEMAH delivery for ALD high-k HfO2 layers