|
SAFC Buchs |
 |
Small Molecule Facilities |
 |
|
| |
|
SAFC Buchs has a long history of producing high-quality intermediates and bulk actives.The plant specializes in complex, multi-step synthesis to 1,600 liters and low temperature reactions (-60°C) up to 400 liters. A broad range of production technologies and multi-purpose equipment offers batch sizes from grams to tons. This facility operates under cGMP, certified by the Swiss authorities.
|
| Services Overview |
| |
Custom synthesis and process development |
| |
Production of high-quality intermediates and bulk actives |
| |
Technology strengths are aimed toward low-volume, highly complicated chemistries |
| |
Reactor capacity from 5 to 400 gallon (20 L - 1,600 L) glass-lined and Hastelloy |
| |
Operating temperature range from -60°C up to +180°C |
| |
Safety of all processes is evaluated using state-of-the-art DSCRC 1 and ARC calorimeter equipment |
| |