| |
|
| |
400 Liter nominal working volume to 720 Liter maximum capacity |
| |
Facilitates fast reaction
time |
| |
Provides a high level of process control
regarding pressure & temperature parameters - cooling rates
of up to 7°C per minute can be achieved |
| |
Ability to re-use catalyst (decant)
for multiple batches |
| |
Sampling possible at high pressure |
| |
Hastelloy C22 |
| |
cGMP Design – has Clean-in-Place (CIP) |
| |
50 barg maximum working pressure |
| |
+15°C to +150°C operating temperature range |
| |