SAFC Sheboygan
Site Overview
Sheboygan Falls, Wisconsin USA
The Sheboygan Production Center is located in Sheboygan Falls, Wisconsin USA and encompasses 427,000 ft2 of manufacturing space. There are 5 production facilities located within the production site that are responsible for the manufacture of over 3,500 organic compounds.
  Core Technologies
Organometallic Reactions 50 – 4,000 GA
Condensation 50 – 4,000 GA
Distillations 50 – 2,000 GA
Comprehensive Expertise Chart
  Core Competencies
Pharmaceutical Intermediates
Air-sensitive Chemicals
Solvents
High Purity Materials
Process Development
 Equipment Summary
Vessels 50–200 GA
(200–800 L)
>200–500 GA
(>800–2,000 L)
>500–2,000 GA
(>2,000–8,000 L)
>2,000 GA
(>8,000 L)
Total
Glass-Lined 9 2 6 1 18
Stainless Steel 3 5 3 0 11
Hastelloy 1 2 1 0 4
Temp. Range (°C) -110 to +250 -110 to +250 -25 to +180 -25 to +180  
Total Capacity 1,500 GA
(6,000 L)
2,300 GA
(9,200 L)
16,000 GA
(64,000 L)
4,000 GA
(16,000 L)
23,800 GA
(95,200 L)
 Production Facilities
Industrial Scale Air-Sensitive Manufacturing 360° Virtual Tour
  Large scale manufacturing of air-sensitive materials and pharmaceutical intermediates.
Intermediate Scale Air-Sensitive Manufacturing 360° Virtual Tour
  Pilot plant for the scale-up and manufacturing of air-sensitive materials and high purity electronics grade chemicals.
Large Scale Organic Manufacturing 360° Virtual Tour
  Large scale synthesis of commercial products and intermediates for the pharmaceutical and electronics industries.
Solvents / High Purity Organics Purification 360° Virtual Tour
  Purification and packaging of solvents and high purity organic liquids.
Process Development 360° Virtual Tour
  Process development and scale-up services for pharmaceutical intermediates and materials science chemicals.
Quality Control 360° Virtual Tour
  Complete laboratories and staff to support all phases of production development and manufacturing.