| |
Catalog Number: |
|
A4268 |
| |
Activity: |
|
≥700 units/mg protein (E1%/280) |
| |
Common Substrate(s) w Product Number(s): |
|
25590 -- maltoheptaoside
25591 -- maltotrioside
N5885 -- maltoside
A4640 -- Amylopectin Azure
A3508 -- Amylose
10140 -- Amylose
85641 -- Starch |
| |
Common Inhibitor (s) w Product Number(s): |
|
D9305 -- 1-deoxynojirimycin
A3535 -- A-amylase inhibitor |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
150,000 mg |
|
|
| |
Catalog Number: |
|
A7011 |
| |
Activity: |
|
≥300 units/mg protein |
| |
Common Substrate(s) w Product Number(s): |
|
459844 -- ethanol
110078 -- acetaldehyde |
| |
Common Inhibitor (s) w Product Number(s): |
|
T1132 -- Tetraethyliuram disulfide |
| |
Cofactor or Coenzyme: |
|
N7004 -- NAD |
| |
Typical Batch Size: |
|
1,500,000 KU |
|
|
| |
Catalog Number: |
|
A6279 |
| |
Activity: |
|
≥3 TIU/mg protein |
| |
Common Substrate(s) w Product Number(s): |
|
Trypsin inhibitor |
| |
Common Inhibitor (s) w Product Number(s): |
|
|
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
200,000 ml |
|
|
| |
Catalog Number: |
|
A6535 |
| |
Activity: |
|
ATPase ≥200 units/mg protein |
| |
Common Substrate(s) w Product Number(s): |
|
Multiple substrates-Some of the more common:
A7699 -- ATP
A2754 -- ADP |
| |
Common Inhibitor (s) w Product Number(s): |
|
33936 -- 1-hydroxy-naphthalene-3,6-disulfonic acid
D3514 -- 4,4'-diisothio-cyanostilbene 2'2'-disulfonic acid |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
1,000,000 units |
|
|
| |
Catalog Number: |
|
G5635 |
| |
Activity: |
|
≥600 units/mg protein |
| |
Common Substrate(s) w Product Number(s): |
|
Multiple substrates-Some of the more common:
67610, 73660, M1633 -- β-D-galactopyranoside
25589 -- β-D-lactoside
69601 -- β-D-galactopyranoside
N1252 -- 4-Nitrophenyl B-D-galactopyranoside
B4387 -- 5-Bromo-3-indolyl β-D-galactopyranoside
16665 -- 5-Bromo-4-chloro-3-indolyl β D-galactopyranoside
F2756 -- Fluorescein di(galactopyranoside) |
| |
Common Inhibitor (s) w Product Number(s): |
|
N2259 1-naphthyl-beta-D-galactoside
A9414, A6560, A9045, A0701 agarose
* beta-galactopyranoside |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
4500 KU |
|
|
| |
Catalog Number: |
|
G0876 |
| |
Activity: |
|
≥85,000 units/mL |
| |
Common Substrate(s) w Product Number(s): |
|
69602 -- 4-Methylumbelliferyl-β-D-glucuronide hydrate
N1875 -- Naphthol AS-BI β-D-glucuronide
77636 -- Phenolphthalein β-D-glucuronide
R2886 -- Resorufin β-D-glucuronide sodium salt |
| |
Common Inhibitor (s) w Product Number(s): |
|
T9165 ψ-Tectorigenin
G7166 D-Glucaro-δ-lactam potassium salt solid
G5269 D-Glucuronic acid |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
20,000 ml |
|
|
| |
Catalog Number: |
|
G7017 |
| |
Activity: |
|
≥100,000 units/mL |
| |
Common Substrate(s) w Product Number(s): |
|
69602 -- 4-Methylumbelliferyl-β-D-glucuronide hydrate
N1875 -- Naphthol AS-BI β-D-glucuronide
77636 -- Phenolphthalein β-D-glucuronide
R2886 -- Resorufin β-D-glucuronide sodium salt |
| |
Common Inhibitor (s) w Product Number(s): |
|
T9165 -- ψ-Tectorigenin
G7166 -- D-Glucaro-δ-lactam potassium salt solid
G5269 -- D-Glucuronic acid |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
20,000 ml |
|
|
| |
Catalog Number: |
|
G8132 |
| |
Activity: |
|
≥1,000,000 units/g solid |
| |
Common Substrate(s) w Product Number(s): |
|
69602 -- 4-Methylumbelliferyl-β-D-glucuronide hydrate
N1875 -- Naphthol AS-BI β-D-glucuronide
77636 -- Phenolphthalein β-D-glucuronide
R2886 -- Resorufin β-D-glucuronide sodium salt |
| |
Common Inhibitor (s) w Product Number(s): |
|
T9165 -- ψ-Tectorigenin
G7166 -- D-Glucaro-δ-lactam potassium salt solid
G5269 -- D-Glucuronic acid |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
800,000 KU |
|
|
| Catalase, from bovine liver |
| |
Catalog Number: |
|
C40 (C9631) |
| |
Activity: |
|
≥10,000 units/mg protein |
| |
Common Substrate(s) w Product Number(s): |
|
Peroxide
R425966 -- 2,2'-azinobis(3-ethylbenzothiazoline-6-sulfonic acid)
D8001 -- 3,3'-diaminobenzidine
A7631 -- ascorbate |
| |
Common Inhibitor (s) w Product Number(s): |
|
A8056 -- 3-Amino-1,2,4-triazole
M6250 -- 2-Mercaptoethanol |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
2,000 g |
|
|
| Deoxyribonuclease I, from bovine pancreas |
| |
Catalog Number: |
|
DN25 (D0876) |
| |
Activity: |
|
Protein ≥85 %, ≥400 Kunitz units/mg protein |
| |
Common Substrate(s) w Product Number(s): |
|
A9272 3'AMP
Double Stranded DNA
circular plasmid DNA |
| |
Common Inhibitor (s) w Product Number(s): |
|
P9375 1,10-Phenanthroline monohydrate
E4378 Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid
E0270 Ethylenediaminetetraacetic acid tripotassium salt dihydrate
N9653 Netropsin dihydrochloride |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
10,000 g |
|
|
| |
Catalog Number: |
|
D5025 |
| |
Activity: |
|
Protein ≥80 %, ≥2,000 Kunitz units/mg protein |
| |
Common Substrate(s) w Product Number(s): |
|
A9272 3'AMP
Double Stranded DNA
circular plasmid DNA |
| |
Common Inhibitor (s) w Product Number(s): |
|
P9375 1,10-Phenanthroline monohydrate
E4378 Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid
E0270 Ethylenediaminetetraacetic acid tripotassium salt dihydrate
N9653 Netropsin dihydrochloride |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
600 g |
|
|
| |
Catalog Number: |
|
E3019 |
| |
Activity: |
|
≥15 units/mg solid |
| |
Common Substrate(s) w Product Number(s): |
|
Multiple substrates-Some of the more common:
N8505 1-Naphthyl acetate
N8000 1-Naphthyl butyrate
I9368 4-Nitrophenyl butyrate
N0758 Naphthol AS-D chloroacetate
83636 Resorufin acetate |
| |
Common Inhibitor (s) w Product Number(s): |
|
T4376 1-chloro-3-tosylamido-4-phenyl-2-butanone
M6250 2-Mercaptoethanol
179124 Acetone
360457 Acetonitrile
E8375 Eserine |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
1,500,000 units |
|
|
| |
Catalog Number: |
|
F4165 |
| |
Activity: |
|
≥0.1 unit/mg solid |
| |
Common Substrate(s) w Product Number(s): |
|
C7078 Casein
F5386 Fibrin |
| |
Common Inhibitor (s) w Product Number(s): |
|
C8917 Cystatin from chicken egg white lyophilized powder
E3132 E-64 |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
1,300,000 units |
|
|
| |
Catalog Number: |
|
G4256 |
| |
Activity: |
|
≥20 units/mg protein (modified Warburg-Christian) |
| |
Common Substrate(s) w Product Number(s): |
|
G0380 alpha-D-galactose 1-phosphate
U1756 UDP-glucose |
| |
Common Inhibitor (s) w Product Number(s): |
|
131377 1,10-Phenanthroline
164984 8-Hydroxyquinoline
A2754 ADP |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
15,000 units |
|
|
| |
Catalog Number: |
|
G6378 |
| |
Activity: |
|
≥200 units/mg protein (modified Warburg-Christian) |
| |
Common Substrate(s) w Product Number(s): |
|
09972 3-Nitrotetrazolium blue chloride
74030, 74032 Nitrotetrazolium blue chloride
88190 Tetrazolium blue chloride |
| |
Common Inhibitor (s) w Product Number(s): |
|
A2056 Acetyl-CoA
A2324, A3650 Amikacin
A2754 ADP
A5394 ATP
N5130 NADPH |
| |
Cofactor or Coenzyme: |
|
N0505 NADP |
| |
Typical Batch Size: |
|
1,500,000 units |
|
|
| |
Catalog Number: |
|
I4504 |
| |
Activity: |
|
>300 units/mg solid |
| |
Common Substrate(s) w Product Number(s): |
|
72555 1-kestose
12255 inulin
L8647 levane |
| |
Common Inhibitor (s) w Product Number(s): |
|
M6250 2-Mercaptoethanol
A6387 Ammonium sulfate
A9880 Aniline
A7506 Ascorbic acid |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
5000 g |
|
|
| |
Catalog Number: |
|
L0382 |
| |
Activity: |
|
Minimum ≥20,000 units.mg protein (using olive oil) |
| |
Common Substrate(s) w Product Number(s): |
|
61710 2-Naphthyl laurate
19362 4-Methylumbelliferyl butyrate
T5069 4-Trifluoromethylumbelliferyl oleate
46942 Fluorescein dibutyrate
57420 Indoxyl acetate
83637 Resorufin butyrate |
| |
Common Inhibitor (s) w Product Number(s): |
|
O4139 Orlistat |
| |
Cofactor or Coenzyme: |
|
|
| |
Typical Batch Size: |
|
250,000, 000 units |
|
|
| |
Catalog Number: |
|
L9506 |
| |
Activity: |
|
≥15 × 106 light units/mg protein |
| |
Common Substrate(s) w Product Number(s): |
|
62645, L6882, 62643 D-Luciferin sodium salt
L9504, L6152 D-Luciferin synthetic
FFW Fireflies, whole
F3641 Firefly Lantern powder
FFT Firefly tails (lanterns)
L9007 Luciferin 6'-ethyl ether
L1147 Luciferin 6'-methyl ether |
| |
Common Inhibitor (s) w Product Number(s): |
|
A9501, 851205 3',5'-cyclic AMP
A6631 Arsenate
126799 Dodecanol
E0270 EDTA |
| |
Cofactor or Coenzyme: |
|
A5394 ATP |
| |
Typical Batch Size: |
|
20,000 mg |