Biocides & Preservatives
For an enviromentally friendly diagnostics manufacturing processes
ProClin® Preservatives:
   Key features of ProClin Preservatives
   Mechanism of Action
   ProClin Evaluation Kits & Packaging
   ProClin Kits' Features
   ProClin Preservatives Product List
ProClin preservatives are an excellent choice for replacing thimerosal, sodium azide and gentamicin preservatives. In addition they are highly effective, broad-spectrum biocides for controlling microorganisms in reagents and products intended for in vitro diagnostics use. At concentrations as low as 0.02%, these preservatives eradicate bacteria, fungi and yeast for extended periods of time.
Register and recieve the new Proclin brochure or efficacy study.
Key features of ProClin preservatives:
Compatibility with key enzymes and assay indicators
Will not inhibit antibody binding
Easy-to-use water-soluble liquid, effective at very low concentrations (6-20ppm)
Excellent stability over a wide pH range (2 to 8.5)
Level of active ingredients assayable by HPLC (Inquire for a HPLC standard)
At recommended usage levels, ProClin presents no health hazards, toxological problems or disposal issues
Mechanism of Action
The active components of ProClin are two isothiazolones, 2-methyl-4-isothiazolin-3-one and 5-chloro-2-methyl-4-isothiazolin-3-one.
Within minutes of contacting a microorganism, the active components penetrate the cell wall and inhibit specific enzymes (at the points noted in red in the Krebs Cycle).
Growth, macromolecule synthesis and respiration are inhibited, ultimately leading to cell death.
Krebs Cycle
ProClin Evaluation Kits
ProClin Evaluation Kits are available for determining the suitability of ProClin products in new product development. Each evaluation kit contains three different manufactured lots of ProClin for testing performance consistency from lot to lot.
To determine the appropriate ProClin product for a specific application, ProClin variety kits are also available. Each kit contains 5 ml of Proclin 150, 200, 300 and 950.
ProClin Evaluation Kits
48121-U ProClin 150 – 3 x 5ml (3 lot numbers)
49381-U ProClin 150 – 3 x 400ml (3 lot numbers)
48911-U ProClin 300 – 3 x 5ml (3 lot numbers)
48119-U ProClin Variety Kit – 5ml each of ProClin 150, 200, 300 & 950 as an aid to select the appropriate ProClin product for a specific application
ProClin Kit Features
  Features ProClin 150 ProClin 200 ProClin 300 ProClin 950
  Active (A.I.) CMIT/MIT CMIT/MIT CMIT/MIT MIT
  % total A.I. 1.5 1.5 3 9.5
  Salt Stabilizers 23 – 25% Mg salts 3% Mg salts Alkyl Carboxylate none
  Matrix Water Water Propylene glycol Water
  Working pH Range 2.5 – 8.5 2.5 – 8.5 2.5 – 8.5 2 – 12
  Typical Dosage Levels 0.05 – 0.10% 0.05 – 0.10% 0.02 – 0.07% 0.05 – 0.10%
  Shelf Life 3 years 3 years 3 years 24 months
ProClin Preservatives Product List
  ProClin Preservatives
  Catalog Number:   Description:
33360-U   ProClin Reference Standard-1ml
ProClin 150    
49376-U   50ml
49377-U   400ml
49378-U   50ml
49379-U   3.6L
49379-U   15L (Pail)
49380-U   110kg (106.8L)
ProClin 200    
48171-U   50ml
500380   400ml
500399   3.6L
500402   15L (Pail)
ProClin 300    
48912-U   50ml
48914-U   400ml
48915-U   2.0L
48916-U   2.8L
48917-U   3.6L
48918-U   18L (Pail)
48919-U   110kg (106.8L)
ProClin 950    
46885-U   5L
46878-U   50L (Pail)
46879-U   400ml
46883-U   3.6L
46884-U   17L (Pail)
Sodium azide    
S8032   Sodium azide
S2002   Sodium azide
71290   Sodium azide
  Catalog Number:   Description:
For further information or to obtain a preservative efficacy study performed by an independent laboratory,
contact your SAFC representative.