Packaging Options
beyond expectation
SAFC Supply Solutions offers a range of customized product packaging services specifically developed to reduce overall process costs, minimize chemical handling and accelerate the route from discovery to commercialization of your products. We have 8 categories of packaging materials with range sizes to assist you with your customized packaging requirements.
 Container Types
Bottles Cans
Ampoules Cylinders
Bags Returnable Container
Boxes Drums
 Accessories
Seals    
Bottles
Container Type Available Sizes
Narrow Mouth Amber Glass Pour Out 4ml - 4000ml
Narrow Mouth Ambert Glass Sure Seal 40ml - 2500ml
Narrow Mouth Amber Glass PVC Coated Sure Seal 125ml - 1000ml
Narrow Mouth Amber Glass PVC Coated 1000ml - 4000ml
Wide Mouth Amber Glass 6cc - 2500cc
Flint Glass 4ml - 2500ml
Narrow Mouth Flint Glass PVC Coated 500ml -2500ml
Wide Mouth Flint Glass 125cc - 4000cc
Narrow Mouth Flint and Amber Glass 4ml - 2500ml
Narrow Mouth Amber Premium HDPE 15ml - 1000ml
Wide MouthAmber Premium HDPE 30ml - 2000ml
Narrow Mouth Natural Premium 15ml - 4000ml
Wide Mouth Natural Premium 30cc - 4000ml
Narrow Mouth Natural Teflon 6ml - 1000ml
Narrow Mouth Aluminum 275ml - 25000ml
Customer Specific 4000ml - 5200ml
Serum Bottles (20mm neck finish) 20mm - 50ml
back to top
Ampoules
Container Type Available Sizes
Amber Glass Ampoules 2ml - 5ml
Flint Glass Ampoules 2ml - 250ml
Flint Glass Funnel Top Ampoules 5ml - 20ml
AAPL Ampoules and Tube Vials 5ml - 200ml
Break Seal Ampoules 5ml - 20ml
NMR Tubes 5ml - 20ml
back to top
Bags
Container Type Available Sizes
Foil Bags 3¼"x6" - 32"x24"
4 mil Polyamide Bags 3¼"x6" - 18"x24"
1½ mil Polyethylene Bags 4"x8" - 12"x24"
1 mil Box Liner - roll 12"x8"x24" - 16"x14"x36"
Zipper Bags 4"x6" - 5"x7"
4 mil Polyethylene Liner Bags 6"x24" - 38"x64"
back to top
Seals
Product Type Available Sizes
Tamper Evident Bands - Cellulose 18x15mm - 85x25mm
Tamper Evident Bands - PVC 18x15mm - 85x25mm
back to top
Boxes
Container Type Available Sizes
CrashLock Boxes for W/M Bottles (solids) 250cc - 1250cc
(for Wide Mouth Amber Glass)
CrashLock Boxes for W/M Bottles (solids) 2000cc - 4000cc
(for Wide Mouth Flint Glass)
CrashLock Boxes for N/M Bottles (liquids) 125ml - 1L
(for Narrow Mouth Amber Glass)
CrashLock Boxes 125ml - 250ml
(for Narrow Mouth Amber Glass)
CrashLock Boxes 250cc - 500cc
(for Wide Mouth Amber Glass)
White Jewlry Boxes (for packaging metals) 4½" - 6½" (with Padded Insert)
Overpack Boxes 6"x6"x9" - 10"x10"x18"
Boxes Case Packs - 3 offers 4x2500ml, 6x1000ml, 6x500ml
Boxes 5"x5"x9" for 125ml N/M Amber Glass Pour Out Bottle
Barrier Packs Consist of Bottle in Bag in Can in Box in Foil Bag in Fiber Overpack Box: See below
Barrier Pack 5"x5"x8" - 8"x8"x10" Fiber Overpack Box; 12"x16" Foil Bag; 3" Silver Lacq. Overpack Can
Barrier Pack for 4000cc Wide Mouth Flint Glass Bottle Fiber Overpack Box; 32"x24" Foil Bag; 8" Gold Lacq. Overpack Can
Barrier Pack 8"x8"x10" - 8"x8"x12" Fiber Overpack Box; 18"x22" Foil Bag; Varied Can sizes
Barrier Pack 7"x7"x12" Fiber Overpack Box; 18"x22" Foil Bag; 6" Silver Lacq. Overpack Can
Barrier Pack - Case Packs 2 Box sizes available
back to top
Cans
Container Type Available Sizes
Paint Cans 500cc - 4000cc
Overpack Cans 3" - 8" Silver lacq. cans & 8" Gold Lacq. Cans
1L Ether Can 1000ml Ether (Safety Can); Pad and Overpack Box (5"x5"x9")
4L Ether Can 4000ml Ether (Safety Can); Pad & Fiber Overpack Box (12"x12"x11")
back to top
Cylinders
Container Type Available Sizes
Low Pressure Carbon Steel Cylinders (Sure-Pac) 25ml - 2250ml
High Pressure 316ss Cylinders (used for Sulfer Trioxide only) 500ml - 2500ml
Returnable Container Single Wall 1000L Single Wall Carbon Steel Tank
Returnable Container Double Wall 400-850L Double Wall Carbon Steel Tank
High Pressure Carbon Steel Cylinders (Lecture Bottles) 440ml bottle;
440ml bottle with stainless steel or brass packless valve
Low Pressure Carbon Steel Cylinders (Kilo-Lab) 8L - 400L
Low Pressure 304ss Cylinders (Pure-Pac) 18L - 400L
Valves ¼" carbon/brass plug - ¼" 304 ss angle packless diaphragm seal valve
Cylinders with Valves 25ml Sure-Pac, with brass or ss plug, and with brass angle needle valve
Cylinders with Valves 280ml Sure-Pac, with brass or carbon steel plug; with steel or brass angle needle valve; bronze plated ball valve
Cylinders with Valves 900ml Sure-Pac, with brass or carbon steel plug; with packless steel angle valve; brass or steel angle needle valve and hex cap
Cylinders with Valves 2400ml 12" cylinders with 3 valve configurations
18L PurePac Cylinder Components Plastic Cylinder Sleeve, Tech Bulletin Al-2-1, 8: sealing wire, SLEFLOK seal w/Red lithod Aldrich, 1-5/8" zinc-placted stell cotter pin, Cylinder Outlet Valve Tag
Low Pressure Carbon Steel Cylinders (Sure-Pac) Packing C Fiber Overpack Box for 4L Bottles,
Top Pad for Sure Pac Cylinder,
2250ml Low Pressure Carbon Steel SurePac Cylinder,
Top Pad for SurePacCylinder
back to top
Drums
Container Type Available Sizes
Open Head Fiber Drums 2½ GA - 55 GA
Closed Head 304ss Drums 7½ GA - 55 GA
Closed Head Steel Drums 10 L - 200 L
Open Head Steel Drums 2 GA - 85 GA
Closed Head Poly Drums 9L (2½ GA) - 55 GA
Closed Head Composite Drums 5 GA - 55 GA
Open Head Poly Drums 7.5L (2 GA) - 18L (5 GA)
back to top