|
Our Offer |
 |
Proligo® Reagents |
 |
|
| |
|
SAFC Supply Solutions manufactures, in the ISO certified Hamburg facility, all raw materials needed for oligonucleotide synthesis: DNA, RNA, 2'O-Methyl, CPG, liquid reagents, modifications and linkers.
SAFC has facilities around the world dedicated to providing services for customers requiring a reliable partner to produce materials and offers: |
|
|
| |
New Products — New Proligo Reagents within our Offer |
| |
Product Breadth — Raw materials for DNA and RNA oligonucleotide synthesis tailored to meet customer specifications |
| |
Custom products — Providing partnerships for custom projects, non-catalogue items are manufactured upon request with predefined cost targets and specifications |
| |
Quality Assurance & Quality Control — High quality and consistency in manufacturing products is routine at SAFC. Our success is driven by one of the industry's leading Quality Management Systems and backed by highly-trained and dedicated QA personnel. Testing flexibility from the industry's leader, we commit to meet the most stringent Quality Control standards to a level personalized for each customer. |
| |
Purification Services — Phosphoramidites are purified at SAFC Hamburg by preparing column chromatography on silica gel with medium to high pressure chromatography equipment |
| |
Packaging Services — Complete packaging services from routine to custom and milligrams to bulk kilogram |
| |
Manufacturing Capacities — After an expansion in 2002, our annual capacity reach 5 metric tons of phosphoramidites production, purification and packaging |