Activator 42™
Proligo® Reagents
Product Breadth of Proligo Reagents
 Phosphoramidites :
Pharmadite®
DNA Phosphoramidites
RNA Phosphoramidites
2'O-Methyl RNA Phosphoramidites
 Fast Deprotection Chemistries :
DNA
RNA
 Supports :
Controlled Pore Glass (CPG)
Columns
 Solvents :
Liquid Reagents
Activator 42™
 Modifications :
Fluorescein Phosphoramidite
Biotin Phosphoramidite
Phosphate-ON Phosphoramidite
Non Standard Nucleosides
 Linkers :
Amino Linkers
ssH Linker
Product Breadth of Proligo Reagents
Activator 42 introduces unprecedented activation power to phosphoramidite coupling reactions. This novel activator provides unique opportunities to speed up oligonucleotide synthesis and to improve on product yield.  
  View Structure:
  •  Activator 42 5-
(Bis-3,5-trifluoro
methylphenyl)-
1H-tetrazole
Pharmadite
Key features of Activator 42
The highest activation efficiency in the industry compared to other activators
Promotes ultrafast DNA couplings (coupling time less than 15 seconds)
Promotes rapid RNA couplings (coupling time 6 minutes)
Excellent solubility in acetonitrile (greater than 0.9M)
Not hygroscopic
Safe to handle (no explosive properties)
Very high overall yield in DNA and RNA synthesis
Solid phase oligonucleotide synthesis with phosphoramidites consists of a sequence of three consecutive reaction steps which are repeated in a cyclical manner, deblocking, coupling and oxidation.  
Whereas deblocking and oxidation reactions proceed with nearly quantitative yields in state of the art oligonucleotide synthesis, the coupling reactions, although high yielding, determine the overall efficiency of the process. Coupling yields are driven by a variety of factors, including amidite and solid support quality, water content of the coupling medium and the choice of activator molecule employed. An ideal activator should promote the coupling of standard DNA amidites as well as the coupling of more sterically demanding amidites (e.g. 2’O-TBDMS-RNA amidites) in highly efficient manner. The activator should have excellent solubility in the coupling medium (i.e. acetonitrile) in order to avoid undesired precipitation or crystallisation, should not be hygroscopic, should be safe to handle and should not lead to side reactions in oligonucleotide synthesis. These desired properties can be attributed by SAFC Proligo Reagents to a new powerful activator molecule which gives superior performance compared to the other commercial activators such as 5-ethylthio-1 H-tetrazole (ETT), 5-benzylthio-1H-tetrazole (BTT) and 4,5-dicyanoimidazole (DCI).
Activation efficiency for 2’O-TBDMS-RNA amidites Activator 42 > ETT = BTT
Activation efficiency for standard DNA amidites Activator 42 > DCI
Product List - Activator 42
Compatible with Expedite™ and Polygen® Instruments
Product No. Description Unit
L0302502-04 Activator 42™ 0.1M 4 x 2.5L
L8300452-06 Activator 42™ 0.1M 6 x 450ml
L8300202-06 Activator 42™ 0.1M 6 x 200ml
L0302512-04 Activator 42™ 0.25M 4 x 2.5L
L8300462-06 Activator 42™ 0.25M 6 x 450ml
L8300212-06 Activator 42™ 0.25M 6 x 200ml
Custom products Other quantities and packaging are available upon request.
SAFC and SAFC Proligo Reagents are trademarks of Sigma-Aldrich Biotechnology, LP.
©2006 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission.
Other trademarks are the property of their respective owners.
To order or speak with a Customer Service Representative, contact your local SAFC office.