RNA Phosphoramidites
Proligo® Reagents
Product Breadth of Proligo Reagents
 Phosphoramidites :
Pharmadite®
DNA Phosphoramidites
RNA Phosphoramidites
2'O-Methyl RNA Phosphoramidites
 Fast Deprotection Chemistries :
DNA
RNA
 Supports :
Controlled Pore Glass (CPG)
Columns
 Solvents :
Liquid Reagents
Activator 42™
 Modifications :
Fluorescein Phosphoramidite
Biotin Phosphoramidite
Phosphate-ON Phosphoramidite
Non Standard Nucleosides
 Linkers :
Amino Linkers
ssH Linker
Product Breadth of Proligo Reagents
RNA Phosphoramidites with Fast Deprotection Chemistry  
  Download Literature:
  •  User Instructions
RNA Phosphoramidites
(770k pdf)
  •  Standard RNA Phosphoramidites
  View Structure:
  •  rA(tac) Phosphoramidite
  •  rC(tac) Phosphoramidite
  •  rG(tac) Phosphoramidite
  •  rU Phosphoramidite
SAFC Proligo Reagents offers RNA phosphoramidite monomers with Fast Deprotection Chemistry (tert-butylphenoxyacetyl (TAC) group).
Advantages of synthesis with TAC-protected RNA phosphoramidites
Increased synthesis yield and reduced purification efforts
Dramatically reduced deprotection times
Minimized exposure to alkaline deprotection medium
Minimized chain degradation during deprotection
RNA Phosphoramidites
Compatible with Expedite™ and Polygen® Instruments
Product No. Description Unit
A211081-01 DMT-2′-O-TBDMS-rA(tac) amidite 1 x 0.5g
C213081-01 DMT-2′-O-TBDMS-rC(tac) amidite 1 x 0.5g
G211081-01 DMT-2′-O-TBDMS-rG(tac) amidite 1 x 0.5g
U211081-01 DMT-2′-O-TBDMS-rU amidite 1 x 0.5g
Compatible with ABI™ Instruments
Product No. Description Unit
A212031-01 DMT-2′-O-TBDMS-rA(tac) amidite 1 x 0.5g
A212061-01 DMT-2′-O-TBDMS-rA(tac) amidite 1 g
C212031-01 DMT-2′-O-TBDMS-rC(tac) amidite 1 x 0.5g
C212061-01 DMT-2′-O-TBDMS-rC(tac) amidite 1 g
G212031-01 DMT-2′-O-TBDMS-rG(tac) amidite 1 x 0.5g
G212061-01 DMT-2′-O-TBDMS-rG(tac) amidite 1 g
U211031-01 DMT-2′-O-TBDMS-rU amidite 1 x 0.5g
U211061-01 DMT-2′-O-TBDMS-rU amidite 1 g
Compatible with MerMade™ Instruments
Product No. Description Unit
A212028-06 DMT-2′-O-TBDMS-rA(tac) amidite 6 x 10g
C212028-06 DMT-2′-O-TBDMS-rC(tac) amidite 6 x 10g
G212028-06 DMT-2′-O-TBDMS-rG(tac) amidite 6 x 10g
U211028-06 DMT-2′-O-TBDMS-rU amidite 6 x 10g
Bulk Quantities
Product No. Description Unit
A212010-01 DMT-2′-O-TBDMS-rA(tac) amidite 1 x 10g
C212010-01 DMT-2′-O-TBDMS-rC(tac) amidite 1 x 10g
G212010-01 DMT-2′-O-TBDMS-rG(tac) amidite 1 x 10g
U211010-01 DMT-2′-O-TBDMS-rU amidite 1 x 10g
Fast Deprotection RNA CPG
Compatible with Expedite™ and Polygen® Instruments
Product No. Description Unit
A602001-01 CPG 500Å, rA(tac), 25-35 μmol/g 1 x 1g
C602001-01 CPG 500Å, rC(tac), 25-35 μmol/g 1 x 1g
G602001-01 CPG 500Å, rG(tac), 25-35 μmol/g 1 x 1g
U601001-01 CPG 500Å, rU, 25-35 μmol/g 1 x 1g
Fast Deprotection RNA Columns
Compatible with Expedite™ Instruments
Product No. Description Unit
A632004-01 Column CPG 500Å, rA(tac), 1 μmol 1 x 4
C632004-01 Column CPG 500Å, rC(tac), 1 μmol 1 x 4
G632004-01 Column CPG 500Å, rG(tac), 1 μmol 1 x 4
U631004-01 Column CPG 500Å, rU, 1 μmol 1 x 4
Compatible with ABI™ Instruments
Product No. Description Unit
A662004-01 Column CPG 500Å, rA(tac), 1 μmol 1 x 4
C662004-01 Column CPG 500Å, rC(tac), 1 μmol 1 x 4
G662004-01 Column CPG 500Å, rG(tac), 1 μmol 1 x 4
U661004-01 Column CPG 500Å, rU, 1 μmol 1 x 4
Custom Products Bulk packaging up to multiple kg per container and customized packaging with alternative quantities of amidites are available upon request.
SAFC and SAFC Proligo Reagents are trademarks of Sigma-Aldrich Biotechnology, LP.
©2007 Sigma-Aldrich Biotechnology LP. All rights reserved. Reproduction forbidden without permission.
Other trademarks are the property of their respective owners.
To order or speak with a Customer Service Representative, contact your local SAFC office.