Analytical / Chromatography > Air Monitoring > Applications > Petrochemical Industry
Air Monitoring Applications
Petrochemical Industry
The petroleum refining industry has advanced from producing cheap kerosene as its main role during inception to its diversity today where the demands of modern transportation have led to production of gasoline, diesel and jet fuels. Industrial innovations have created the demand for petrochemical products such as lubricants, solvents, process oils and resins; also, specialty products like asphalt, wax and coke. Modern oil refineries produce a variety of different products including many that are used in the petrochemical industry as feedstock.
Employees who work in this industry are at risk for exposure to chemical compounds detrimental to their health and safety. Communities may also assume similar risks. Supelco offers a range of products for air monitoring of industrial conditions to assure worker and community safety.
Our air monitoring products are useful for sampling of Indoor Air and Ambient Air.
Contaminants of Interest for the Petrochemical Industry
AIr Monitoring Petrochemical Applications
Air Sampling Media by Regulatory Method
Method Contaminants of Interest Sampling Media
OSHA
OSHA 7 Organic Vapors Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U)
Passive Sampling – Radiello BTEX/VOCs (RAD130)
OSHA 12 Benzene Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U)
Passive Sampling – Radiello BTEX/VOC (RAD130)
OSHA 58 Coal Tar Pitch Volatiles & Selected PAHs Filter – A/E Glass Fiber Filter, 1mm, 37 mm (23378)
OSHA 1002 Xylenes / Ethylbenzene Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U)
OSHA 1005 Benzene Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U)
Passive Sampling – Radiello BTEX/VOCs (RAD130)
Passive Sampling – Radiello BTEX/VOCs (RAD130 / RAD145)
OSHA 1008 Hydrogen Sulfide Sorbent Tube w/Reservoir – Silver Nitrate Coated Silica Gel +
  Sodium Carbonate/Glycerol coated GFF, PUF, SGW Plugs (New)
Passive Sampling – Radiello H2S (RAD170)
OSHA 1011 Sulfur Dioxide Sorbent Tube w/Reservoir – Silver Nitrate Coated Silica Gel +
  Sodium Carbonate/Glycerol coated GFF, PUF, SGW Plugs (New)
Passive Sampling – Radiello HF/NO2/SO2 (RAD166)
OSHA ID 110 Fluorides (F and HF) Filter – Mixed Cellulose Ester (MCE), GN-4 Metricel, 37 mm (23381-U)
Passive Sampling – Radiello HF/NO2/SO2 (RAD166)
OSHA ID 111 Phosphoric Acid Filter – Mixed Cellulose Ester (MCE), GN-4 Metricel, 37 mm (23381)
OSHA ID 113 Sulfuric Acid Filter – Mixed Cellulose Ester (MCE), GN-4 Metricel, 37 mm (23381)
OSHA ID 145 Particulate Mercury Filter – Mixed Cellulose Ester (MCE), GN-4 Metricel, 37 mm (23381)
NIOSH
NIOSH 1501 Aromatic Hydrocarbons Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U)
Passive Sampling – Radiello BTEX/VOCs (RAD130)
NIOSH 1615 Methyl tert-Butyl Ether (MTBE) Sorbent Tube – Coconut Charcoal; ORBO-32L (20228)
NIOSH 3700 Benzene by Portable GC Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U)
Passive Sampling-Radiello BTEX/VOCs (RAD130)
NIOSH 5506 Polynuclear Aromatic Hydrocarbons (PAHs) by HPLC Filter – Zefluor™ PTFE membrane, 2 mm, 37 mm (23390-U),
Sorbent Tube – XAD-2; ORBO-43 (20258)
NIOSH 5515 Polynuclear Aromatic Hydrocarbons (PAHs) by GC Filter – Zefluor™ PTFE membrane, 2 mm, 37 mm (23390-U),
Sorbent Tube – XAD-2; ORBO-43 (20258)
NIOSH 5042 Benzene-Soluble Fraction & Total Particulate (Asphalt Fume) Filter – PTFE, 5 mm, 37mm (23383), Cassette (23370-U), Support Pad (23385)
NIOSH 6013 Hydrogen Sulfide Filter – Zefluor™ PTFE membrane, 0.5 mm, 25 mm (23395)
Sorbent Tube – Coconut Charcoal; ORBO-32L (20228)
Passive Sampling – Radiello H2S (RAD170)
NIOSH 7903 Inorganic Acids Sorbent Tube – Silica GFF; ORBO-53 (20265)
ASTM
ASTM D3686 Organic Vapors Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U)
Passive Sampling – Radiello BTEX/VOCs (RAD130)
ASTM D6196 Volatile Organic Compounds (VOC’s) Thermal Desorption Tubes
Passive Sampling – Radiello BTEX/VOCs (RAD145)
ASTM D6209 Polynuclear Aromatic Hydrocarbons (PAHs) PUF Samplers w/XAD-2 (21233-U, 21235-U)
EPA
EPA TO-13A Polynuclear Aromatic Hydrocarbons (PAHs) by GC/MS Polyurethane Foam (PUF); ORBO-2000 (20037)
EPA IP-7 Determination of benzo(a)pyrene and other Polynuclear Aromatic Hydrocarbons (PAHs) Polyurethane Foam (PUF); ORBO-2000 (20037)
EPA TO-17 Polar and non-polar VOC’s, select aldehydes Sorbent Tube – Solvent Desorption & DNPH Tubes,
   Thermal Desorption TD Tubes
Passive Sampling – Radiello BTEX/VOCs (RAD130 / RAD145)
ORBO-65P (20029-U), 65M (20028-U)
Analytical Reference Standards by Regulatory Agency
View the complete listing of available chemical standards grouped by regulatory agency in our web catalog.
Or, use our Standards Explorer to perform a quick search.
Application Chromatograms
View GC applications for volatiles, hydrocarbons, and semivolatiles
View Air Monitoring applications