| Air Sampling Media by Regulatory Method |
| Method |
Contaminants of Interest |
Sampling Media |
| OSHA |
| OSHA 7 |
Organic Vapors |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) Passive Sampling – Radiello BTEX/VOCs (RAD130) |
| OSHA 12 |
Benzene |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) Passive Sampling – Radiello BTEX/VOC (RAD130) |
| OSHA 48 |
Petroleum Distillates |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) |
| OSHA 58 |
Coal Tar Pitch Volatiles & Selected PAHs |
Filter – A/E Glass Fiber Filter, 1mm, 37 mm (23378) |
| OSHA 83 |
2-Butoxyethanol |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) Passive Sampling – Radiello BTEX/VOC (RAD130) |
| OSHA 111 |
Toluene |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) Passive Sampling – Radiello BTEX/VOCs (RAD130 / RAD145) |
| OSHA 1002 |
Xylenes / Ethylbenzene |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) Passive Sampling – Radiello BTEX/VOCs (RAD130 / RAD145) |
| OSHA 1005 |
Benzene |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) Passive Sampling – Radiello BTEX/VOCs (RAD130 / RAD145) |
| OSHA 1007 |
Formaldehyde |
Passive Sampling – Radiello DNPH (RAD165) |
| OSHA 1008 |
Hydrogen Sulfide |
Sorbent Tube w/Reservoir – Silver Nitrate Coated Silica Gel + Sodium Carbonate/Glycerol coated GFF, PUF, SGW Plugs (New) Passive Sampling – Radiello H2S (RAD170) |
| OSHA 1011 |
Sulfur Dioxide |
Sorbent Tube w/Reservoir – Silver Nitrate Coated Silica Gel + Sodium Carbonate/Glycerol coated GFF, PUF, SGW Plugs (New) Passive Sampling – Radiello HF/NO2/SO2 (RAD166) |
| OSHA ID 110 |
Fluorides (F and HF) |
Filter – Mixed Cellulose Ester (MCE), GN-4 Metricel, 37 mm (23381-U) Passive Sampling – Radiello HF/NO2/SO2 (RAD166) |
| OSHA ID 111 |
Phosphoric Acid |
Filter – Mixed Cellulose Ester (MCE), GN-4 Metricel, 37 mm (23381) |
| OSHA ID 113 |
Sulfuric Acid |
Filter – Mixed Cellulose Ester (MCE), GN-4 Metricel, 37 mm (23381) |
| OSHA ID 145 |
Particulate Mercury |
Filter – Mixed Cellulose Ester (MCE), GN-4 Metricel, 37 mm (23381) |
| OSHA PV2051 |
Propylene Glycol |
OSHA Versatile Sampler –OVS-7 (Custom-Inquire) |
| OSHA 165SG |
Acid Gases and Mists |
Sorbent Tube – Silica Gel w/GFF ORBO-53 (20265) |
| NIOSH |
| NIOSH 1501 |
Aromatic Hydrocarbons |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) Passive Sampling – Radiello BTEX/VOCs (RAD130) |
| NIOSH 1615 |
Methyl tert-Butyl Ether (MTBE) |
Sorbent Tube – Coconut Charcoal; ORBO-32L (20228) |
| NIOSH 2541 |
Formaldehyde by GC |
Sorbent Tube – 20HMP Coated XAD-2; ORBO-23 (20257) Passive Sampler – Radiello Aldehydes (RAD165) |
| NIOSH 3700 |
Benzene by Portable GC |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) Passive Sampling-Radiello BTEX/VOCs (RAD130) |
| NIOSH 5506 |
Polynuclear Aromatic Hydrocarbons (PAHs) by HPLC |
Filter – Zefluor™ PTFE membrane, 2 mm, 37 mm (23390-U), Sorbent Tube – XAD-2; ORBO-43 (20258) |
| NIOSH 5515 |
Polynuclear Aromatic Hydrocarbons (PAHs) by GC |
Filter – Zefluor™ PTFE membrane, 2 mm, 37 mm (23390-U), Sorbent Tube – XAD-2; ORBO-43 (20258) |
| NIOSH 5042 |
Benzene-Soluble Fraction & Total Particulate (Asphalt Fume) |
Filter – PTFE, 5 mm, 37mm (23383), Cassette (23370-U), Support Pad (23385) |
| NIOSH 5800 |
Polyaromatic Compounds (PACs), Total |
Sorbent Tube – XAD-2; ORBO-43 (20258) |
| NIOSH 6013 |
Hydrogen Sulfide |
Filter – Zefluor™ PTFE membrane, 0.5 mm, 25 mm (23395) Sorbent Tube – Coconut Charcoal; ORBO-32L (20228) Passive Sampling – Radiello H2S (RAD170) |
| NIOSH 7903 |
Inorganic Acids |
Sorbent Tube – Silica GFF; ORBO-53 (20265) |
| ASTM |
| ASTM D3686 |
Organic Vapors |
Sorbent Tube – Coconut Charcoal; ORBO-32S (20267-U) Passive Sampling – Radiello BTEX/VOCs (RAD130) |
| ASTM D6196 |
Volatile Organic Compounds (VOCs) |
Thermal Desorption Tubes Passive Sampling – Radiello BTEX/VOCs (RAD145) |
| ASTM D6209 |
Polynuclear Aromatic Hydrocarbons (PAHs) |
PUF Samplers w/XAD-2 (21233-U, 21235-U) |
| EPA |
| EPA TO-13A |
Polynuclear Aromatic Hydrocarbons (PAHs) by GC/MS |
Polyurethane Foam (PUF); ORBO-2000 (20037) |
| EPA IP-7 |
Determination of benzo(a)pyrene and other Polynuclear Aromatic Hydrocarbons (PAHs) |
Polyurethane Foam (PUF); ORBO-2000 (20037) |
| EPA TO-15 |
Toxic Organic Compounds in Ambient Air |
Thermal Desorption Tubes Passive Sampling – Radiello BTEX/VOCs (RAD130 / RAD145) |
| EPA TO-17 |
Polar and non-polar VOCs, select aldehydes |
Sorbent Tube – Solvent Desorption & DNPH Tubes, Thermal Desorption Tubes Passive Sampling – Radiello BTEX/VOCs (RAD130 / RAD145) ORBO-65P (20029-U), 65M (20028-U) |