|
Air Monitoring
Adsorbent Selection
|
|
|
In addition to the general guidelines presented in the tables below view the technical report titled “A Tool for Selecting an Adsorbent for Thermal Desorption Applications” (1,770Kb pdf - T402025), which highlights the relative differences between the most commonly used adsorbents with the help of color-coded performance charts.
| General Guidelines for Choosing Carbon Adsorbents |
back to top |
For multi-bed tubes, use the weaker adsorbent in front of the stronger adsorbent. For example, use Carbopack™ C in front of Carbopack B.
| Relative Analyte Size (1) |
Recommended Materials (listed weakest to strongest) |
| > C20 |
Carbotrap™ F, Carbopack F |
| C12-C20 |
Carbotrap C, Carbopack C, Carbotrap Y, Carbopack Y |
| C5-C12 |
Carbotrap B, Carbopack B |
| C3-C9 |
Carboxen™ 1016, Carbotrap X, Carbopack X, Carbopack Z |
| C2-C5 |
Carboxen 569, Carbosieve™ G, Carboxen 1000, Carbosieve S-III, Carboxen 1021, Carboxen 1018, Carboxen 1003, Carboxen 1012 |
| (1) Analyte size relative to n-Alkanes. Consider all atoms, not just Carbon. For example, even though 1,2-Dichloroethane is a C2, the two Chlorine atoms give it a relative size between C4 and C5. |
| Physical Characteristics of Supelco Carbon Adsorbents |
back to top |
|
| Adsorbent |
BET surface area1 (m2/g) |
Density (g/mL) |
Porosity (cc/g) |
Micropore Diameter (Å) |
| micro- |
meso- |
macro- |
| Carbotrap graphitized carbon black (20/40 mesh ) |
| Carbotrap F |
5 |
0.69 |
- |
- |
- |
- |
| Carbotrap C |
10 |
0.68 |
- |
- |
- |
- |
| Carbotrap Y |
24 |
0.45 |
- |
- |
- |
- |
| Carbotrap B |
100 |
0.37 |
- |
- |
- |
- |
| Carbotrap X |
240 |
0.43 |
- |
0.62 |
- |
100 |
| Carbopack graphitized carbon black (60/80 mesh) |
| Carbopack F |
5 |
0.64 |
- |
- |
- |
- |
| Carbopack C |
10 |
0.68 |
- |
- |
- |
- |
| Carbopack Y |
24 |
0.42 |
- |
- |
- |
- |
| Carbopack B |
100 |
0.35 |
- |
- |
- |
- |
| Carbopack Z |
220 |
0.18 |
- |
1.73 |
- |
255 |
| Carbopack X |
240 |
0.41 |
- |
0.62 |
- |
100 |
| Carbon Molecular Sieve |
| Carboxen-1016 |
75 |
0.40 |
- |
0.34 |
- |
- |
| Carboxen-569 |
485 |
0.58 |
0.20 |
0.14 |
0.10 |
5 - 8 |
| Carboxen- 10212 |
600 |
0.62 |
0.30 |
- |
- |
5 - 8 |
| Carboxen-10182 |
675 |
0.60 |
0.35 |
- |
- |
6 - 8 |
| Carbosieve S-III3 |
975 |
0.61 |
0.35 |
0.04 |
- |
4 - 11 |
| Carboxen-1003 |
1000 |
0.46 |
0.38 |
0.26 |
0.28 |
5 - 8 |
| Carbosieve G |
1160 |
- |
0.49 |
0.02 |
- |
6 - 15 |
| Carboxen-1000 |
1200 |
0.48 |
0.44 |
0.16 |
0.25 |
10 - 12 |
| Carboxen-1012 |
1500 |
0.50 |
- |
0.66 |
- |
19 - 21 |
1 Brunauer, Emmett, Teller (BET) surface area calculations 2 microporous, monoporous carbon sieve 3 closed pore structure |
|
|
| Adsorbent |
Supelco Equivalent or Substitute |
| Ambersorb® 347 |
Carboxen 564 |
| Anasorb C300 |
Hopcalite |
| Anasorb CMS |
Carboxen 564, Carboxen 1000, Carbosieve S-III |
| Anasorb CSC |
Coconut charcoal |
| Anasorb GCB1 |
Carbopack B, Carbotrap B |
| Anasorb GCB2 |
Carbopack C, Carbotrap C |
| Anasorb 708 |
Chromosorb® 108 |
| Anasorb 727 |
XAD-4 |
| Anasorb 747 |
Carboxen 564, Carboxen 1000, Carbosieve S-III |
| Carbograph 1 |
Carbopack B, Carbotrap B |
| Carbograph 2 |
Carbopack C, Carbotrap C |
| Carbosphere |
Carboxen 1000 |
| Graphpac-GC |
Carbopack B, Carbotrap B, Carbopack C Carbotrap C |
| Hydrar |
Hopcalite |
| Purosieve |
Carboxen 1000 |
| Spherocarb |
Carboxen 1000 |
| Tenax® GC |
Tenax® TA |