Analytical / Chromatography > Air Monitoring > radiello Diffusive Sampling
Analytical / Chromatography
radiello Diffusive Sampling
radiello® passive/diffusive samplers for environmental monitoring, ambient and outdoor air monitoring, industrial hygiene, industrial air quality (IAQ), personal sampling and breathing zone assessment. The radial design offers higher capacity and faster uptake/sampling rates than traditional passive monitors. Applications include BTEX, VOCs, NO2, SO2, HF, aldehydes, ozone, H2S, ammonia, HCl, phenols, anesthetic gases/vapors and 1,3-butadiene & Isoprene.
Products:
Special Offers:
Toolbox
Learning Center:
Related Topics:
Information & News
Radiello 1,3-Butadiene Sampler
Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
radiello brochure
New! icon radiello® brochure (request)
Supelco Fluka Analytical Catalogs
Supelco Catalog,
Fluka Reagents & Standards Catalog
sign-up for newsletters
Sign-up for newsletters
& e-newsletters!