Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Analytical / Chromatography > Air Monitoring > radiello Diffusive Sampling > Learning Center > Radiello® 1,3-Butadiene Diffusive Sampler for measuring workplace atmospheres
Radiello Diffusive Sampling
Radiello® 1,3-Butadiene Diffusive Sampler for measuring workplace atmospheres
CarbopackDiffusive SamplerThe manufacturing of elastomers makes wide use of 1,3-butadiene (CAS no. 106-99-0) and isoprene (2-methyl-1,3-butadiene, CAS no. 78-79-5). Both compounds are classified as carcinogenic by the International Agency for Research on Cancer (IARC). 1,3 butadiene is listed in group 1, carcinogenic for humans, and isoprene is in group 2B, possibly carcinogenic for humans. The measurement of these compounds in workplace air requires therefore high sensitivity and also the control of explosive hazards, which are frequent in chemical plants.
The diffusive sampling followed by thermal desorption and GC-MS analysis with the new Radiello Butadiene sampler represents a simple and affordable tool for this measurement, while satisfying the above requirements.
Carbopack
The sampler consists of a cartridge adsorbent filled with Carbopack™ X, a graphitised carbon black developed and manufactured by Supelco, that is placed in the Radiello yellow diffusive body (RAD1202). After sampling the cartridge adsorbent is placed in a TD empty tube and analyzed by thermal desorption (TD)/GC-MS.
The nominal sampling rate (@20°C, 50% RH, for 8h exposure) for 1,3-Butadiene is 30,5 +/- 0,3 ml/min 34-2960 µg/m3 , for Isoprene 
41,2 +/- 4,9 mL/min @ 2-6600µg/m3.
Ordering Information
Cat. No. Description PK. Add to Cart
RAD141 Cartridge Adsorbent 1,3-Butadiene/Isoprene 20
RAD121 Triangular Support Plate 20
RAD1202 Diffusive Body, Yellow 20
RAD122 Vertical Adapter for Personal Sampling 20
RAD141S Starter Kit 1,3-Butadiene/Isoprene Sampler
- 1 support plate,
- 1 yellow diffusive body,
- 1 vertical Adapter,
- 2 cartridge adsorbent w/Carbopack X
1
Technical Resources / Literature
Data Sheet for RAD141 (388 KB PDF)
Data Sheet for RAD141S Starter Kit (1 MB PDF)
Field Validation of Radiello Butadiene sampler vs. OSHA 56 – Summary (195 KB PDF)
Supelco Reporter 37 Europe - Radiello Butadiene Article (199 KB PDF)
Cartridge Adsorbent
Carbopack
Starter Kit