To conduct air sampling using the radiello system, the minimum requirements are: cartridge adsorbent, diffusive body, adhesive labels for tracking sampling date/time, and a support plate for attaching the diffusive body-cartridge assembly.
Cartridge AdsorbentsDiffusive Bodies & Support plateStarter Kits for VOC/BTEXReady-to-use SamplerAccessories and SparepartsCalibration Solultion and Kits
Back to radiello diffusive sampling Home