|
To conduct air sampling using the radiello system, the minimum requirements are: cartridge adsorbent, diffusive body, adhesive labels for tracking sampling date/time, and a support plate for attaching the diffusive body-cartridge assembly.
Cartridge Adsorbents Diffusive Bodies & Support plate Starter Kits for VOC/BTEX Ready-to-use Sampler Accessories and Spareparts Calibration Solultion and Kits
Back to radiello diffusive sampling Home
|