Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
radiello Products & Accessories
Accessories & Spare Parts
Outdoor Shelter
On-Field Thermometer and Reader
radiello Barcode Labels
Empty Storage Tubes
Outdoor Shelter
Polypropylene protective outdoor shelter for radiello samplers
A polypropylene protective outdoor shelter for housing up to four radiello samplers during outdoor/ambient air sampling. The shelter allows for adequate ventilation while simultaneously protecting the samplers from harsh weather conditions. The shelter can be mounted to a variety of street fixtures including lampposts, and traffic light and telephone poles of various diameters. The shelter can be easily mounted without the use of tools. The shelter comes in a pack of 10 and includes 20 mounting strips.
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
196
Outdoor Shelter
10
198
Spare Mounting Strips
100
On-Field Thermometer and Reader
Uptake rates are dependent on temperature; therefore, concentration values obtained during sampling will be more accurate if precise temperature values are recorded during sampling. Note that temperature variations of 4-5°C can be recorded from one area of an urban population to another.
The radiello thermometer acts like a mini (< 1 cm3) temperature measurement station that can be mounted on the triangular support plate in conjunction with the diffusive body via a pre-attached vertical adapter. The thermometer offers a precision of ± 0.5°C between –20 to 80°C, and can log up to 2048 temperature data points allowing you to record one temperature reading every 15 min. for 22 days, every 30 min. for 43 days, or 60 minutes for 85 days. The thermometer requires no batteries, and is amenable to harsh weather conditions.
A temperature reader (purchased separately) connects the thermometer to your PC (via RS232 serial port) allowing the user to program the thermometer before sampling and download temperature readings after sampling. Each thermometer has a unique serial number for easy identification. A user-friendly software package is included with the reader to perform statistical and graphic analysis.
radiello thermometer
Reader (RAD127)
Statistical/Graphic Software
Thermometer
Reader (RAD127)
Statistical/Graphic Software
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
126
radiello Thermometer
3
126-1
radiello Thermometer for ready-to-use sampler
3
127
radiello Thermometer Reader
1
radiello Barcode Labels
radiello Barcode Labels
Each label is self-adhesive with a unique barcode for the unmistakable identification of the cartridge adsorbent during sampling, desorption, and analysis.
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
190
radiello Barcode Labels
198
Empty Storage Tubes
radiello RAD1991
radiello RAD1992
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
199-1
radiello Glass Tube, 2.8 mL with stopper
20
199-2
radiello PP Tube, 12 mL, with stopper
20
Back to Top
Back to radiello Products & Accessories
Back to radiello diffusive sampling Home