Sigma-Aldrich
Analytical / Chromatography > Air Monitoring > radiello Diffusive Sampling > Products & Accessories > Cartridge Adsorbents
Products Accessories
Cartridge Adsorbents
Several different cartridge adsorbents are available. Each of which are specific for different classes of compounds. The dimensions for each cartridge are 60 mm L x 4.8 or 5.8 mm diameter. Each cartridge is designed for one time use with the exception of Thermal Desorption (TD) cartridge adsorbents
A sufficient number of adhesive labels with barcode is included with every pack of cartridges for easy tracking of sampling date and time.
cartridge_adsorbents
Compound Class
radiello Code
Description of adsorber bed
Pkg Qty.
Supelco Cat. No.
Add to cart
VOCs, and BTEX
CS2 Desorption
130
SS net (5.8mm diam), activated charcoal
20
RAD130
VOCs and BTEX
Thermo Desorption
145
SS net (4.8mm diam), Carbograph 4
20
RAD145
1,3-Butadiene/Isoprene
141
SS net (4,8mm diam.), Carbopack X
20
RAD141
Aldehydes
165
SS net w/ 2,4-DNPH coated florisil
20
RAD165
Ammonia
168
Microporous PE impregnated with phosphoric acid
20
RAD168
Anaesthetic gases/vapors
132
SS net w/ mix of mol sieve & activated charcoal
20
RAD132
HCl
169
SS net w/silica gel
20
RAD169
HF, NO2 & SO2
166
Microporous PE coated w/ wet TEA
20
RAD166
Hydrogen Sulfide
170
Microporous PE impregnated with zinc acetate
20
RAD170
Ozone
172
Micropore PE tube with silica coated with 4,4'-dipyrldylethylene
20
RAD172
Phenolic Compounds
Thermo Desorption
147
SS net (4.8mm diam),Tenax-TA
20
RAD147
Back to radiello Products & Accessories
Back to radiello diffusive sampling Home