Decrease Font Size Increase Font Size Email this page to a friend Printer Friendly Page
Products & Accessories
Diffusive Bodies
Diffusive Bodies
Triangular Support Plate
Vertical Adapter
Diffusive Bodies
diffusive bodies The diffusive bodies are designed to house radiello cartridge adsorbents during sampling. Dimensions are 60 mm height x 16 mm diameter.
There are 4 different diffusive bodies available. Each of which are application specific in design. Please refer to the radiello Manual or use the listing of the ready-to-use sampler for selection.
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
120
White Diffusive Body, microporous PE, equivalent diffusive path length 18 mm
20
120-1
Blue Diffusive Body, same specifications as White Diffusive Body but darkened with blue dye to protect the cartridge from sunlight
20
120-2
Yellow Diffusive Body, microporous PE (smaller pores) equivalent diffusive path length 150 mm
20
120-3
Permeative Diffusive Body, silicone membrane with SS net
20
Triangular Support Plate
triangular support plate The triangular support plate acts as both a closure and means of suspension for the diffusive body and cartridge adsorbent during sampling. Each support plate is threaded for easy diffusive body attachment. Each plate includes a clip and transparent adhesive pocket to hold the barcode label.
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
121
Triangular Support Plate
20
195
Replacement clips for code 121
20
Vertical Adapter
For personal sampling (breathing zone assessment), a vertical adapter is available to position the diffusive body vertically on the triangular support plate.
vertical adapter
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
122
Vertical Adapter
20
122-1
Snapring adapters for ready to use samplers
20
Back to Top
Back to radiello Products & Accessories
Back to radiello diffusive sampling Home