Sigma-Aldrich
Analytical / Chromatography > Air Monitoring > radiello Diffusive Sampling > Products & Accessories > Ready-to-use Samplers
radiello® Products & Accessories
Ready-to-use Samplers
The radiello ready-to-use diffusive samplers come pre-assembled with the cartridge adsorbent pre-contained within the diffusive body, and sealed with a polycarbonate screw-thread cap. To avoid premature sampling, the entire unit (diffusive body with pre-sealed and installed cartridge adsorbent) is enclosed in an airtight polypropylene (PP) container. Just before use, the unit is removed from the PP container, and snapped into the ready-to-use vertical adapter pre-fixed to the triangular support plate. Once sampling is complete, the diffusive sampling unit is removed from the support plate and resealed into the PP container for shipment to the lab.
diffusive_samplers
Each ready-to-use sampler includes: a sampler unit (sealed diffusive body with cartridge adsorbent), glass or plastic tube, ready-to-use vertical adapter, and barcode label and PP container. Please order the triangular support plates (RAD121) separately.
Note that the ready-to-use diffusive samplers are ideal for workplace sampling campaigns, but are not for sampling low concentrations in outdoor or domestic environments. The shelf life of these samplers is 3 months.
Application
radiello Code
Description
Pkg Qty.
Supelco
Cat. No.
Add to Cart
VOCs and BTEX
CS2 Desorption
123-1
Code 120 White Diffusive Body (Cat. No. RAD120) and Code 130 Cartridge Adsorbent (Cat. No. RAD130)
5
RAD1231
VOCs and BTEX
Thermo Desorption
123-2
Code 120-2 Yellow Diffusive Body (Cat. No. RAD1202) and Code 145 (Cat. No. RAD145) Cartridge Adsorbent
5
RAD1232
HF, NO2 & SO2
123-3
Code 120-1 Blue Diffusive Body (Cat. No. RAD1201) and Code 166 Cartridge Adsorbent (Cat. No. RAD166)
5
RAD1233
Aldehydes
123-4
Code 120-1 Blue Diffusive Body (Cat. No. RAD1201) and Code 165 Cartridge Adsorbent (Cat. No. RAD165)
5
RAD1234
Ozone
123-5
Code 120-1 Blue Diffusive Body (Cat. No. RAD1201) and Code 172 Cartridge Adsorbent (Cat. No. RAD172)
5
RAD1235
Hydrogen Sulfide
123-6
Code 120 White Diffusive Body (Cat. No. RAD120) and Code 170 Cartridge Adsorbent (Cat. No. RAD170)
5
RAD1236
Ammonia
123-7
Code 120-1 Blue Diffusive Body (Cat. No. RAD1201) and Code 168 Cartridge Adsorbent (Cat. No. RAD168)
5
RAD1237
HCl
123-8
Code 120 White Diffusive Body (Cat. No. RAD120) and Code 169 Cartridge Adsorbent (Cat. No. RAD169)
5
RAD1238
Anesthetic Gases & Vapors
125
Includes: 1 permeative body (like RAD1203), 1 support plate (like RAD121), 1 vertical adapter (like RAD122), 1 adsorbing cartridge (like RAD132)
10
RAD125
Back to radiello Products & Accessories
Back to radiello diffusive sampling Home
<