|
Air Monitoring
Solvent Desorption Tubes
|
|
|
LpDNPH cartridges are air-sampling devices designated for sampling carbonyls (e.g. formaldehyde) in ambient, indoor, and industrial atmospheres. Carbonyls are trapped on a high purity silica adsorbent coated with 2,4-dinitrophenylhydrazine,where they are converted to hydrazone derivatives. The derivatives are eluted from the cartridge with acetonitrile and analyzed by HPLC.
Features & Benefits of LpDNPH cartridges:
- High purity adsorbent for better accuracy and trace analysis.
- High sensitivity, low background.
- Low pressure drop, enables use at high sampling rates (1.5-2 L/min)
- New packaging, for maximum sample integrity
- Manufactured in controlled environment
- Meet EPA, OSHA, NIOSH and ASTM specifications
|
| LpDNPH Cartridge Specifications |
| Adsorbent: |
Chromatographic grade, high purity silica gel coated with: DNPH: 2,4-dinitrophenylhydrazine BPE: trans-1,2-bis(4-pyridyl)ethylene |
| Particle Size: |
150-250µm (60/100 mesh) |
| DNPH Loading: |
0.29% (1mg/cartridge) 0.86% (3mg/cartridge, H series) 0.21% (0.25mg/cartridge, ORBO™-DNPH) |
| Bed Weight: |
350mg (S10 series) 350mg (H10) 120mg (ORBO™-DNPH) 1g (H30) 10g (H300) |
| Capacity (total carbonyls): |
75µg (S10 series) 225µg (H10) 643µg (H30) 6400µg (H300) 18.8µg (ORBO™-DNPH) |
| Cartridge Length: |
7.4cm (S10) 3.8cm (S10x) 4.0cm (S10L) 4.0cm (Rezorian™) 9.0cm (ORBO™-DNPH) |
| Background: |
SPE Style Cartridges <0.06µg formaldehyde <0.10µg acetaldehyde <0.30µg acetone |
ORBO™-DNPH <0.025µg formaldehyde <0.035µg acetaldehyde <0.120µg acetone |
| Pressure Drop: |
<7pKa at 1.5L/min <20 inches water at 167mL/min (<28 inches water/<2.1 inches mercury) |
| Agency Method Applications |
| |
US EPA TO-11A-Method for Determination of Formaldehyde in Ambient Air Using Adsorbent Cartridge Followed by High Performance Liquid Chromatography (HPLC) |
| |
US EPA IP-6A-Determination of Formaldehyde and Other Aldehydes in Indoor Air Using a Solid Sorbent Cartridge |
| |
US EPA-Technical Assistance Document for Sampling and Analysis of Ozone Precursors |
| |
ASTM D5197-Standard Test Method for Determination of Formaldehyde and Other Carbonyl Compounds in Air (Active Sampler Methodology |
|