|
AQUANALTM
Analytical Case for Building Restoration
|
Concept
 |
The analytical case for building restoration is designed for investigating the efflorescing of masonry, for aiding decontamination / redevelopment work and for analysing water and etravasted substances in soil in respect of constituents harmful to buildings.
The case contains tests for the determination of chloride, total hardness, magnesium, nitrate, sulfate and sulfide.
The operating manual gives advise how to prepare samples and shows how to calculate the results.
The evaluation of the results can either be done with Spectro 1 photometer or with Spectro 2 photometer - the photometers must be orded seperately!
|
Ordering Information
| Product No. |
Product Name |
Add to Cart
|
| 37563 |
AQUANALTM-plus analytical case for building restoration |
|
| 37560 |
Spectro 1 photometer for AQUANALTM-plus-reagent |
|
| 37781 |
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue German |
|
| 37782 |
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue English |
|
| 37783 |
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue French |
|
| 37784 |
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue Italian |
|
| 37785 |
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue Spanish |
|
| 37441 |
refill pack chloride reagents |
|
| 37456 |
refill pack total hardness reagents |
|
| 37466 |
refill pack magnesium reagents |
|
| 37449 |
refill pack nitrate reagents |
|
| 37469 |
refill pack sulfate reagents |
|
| 37470 |
refill pack sulfide reagents |
|
Back to top Compact Labs home AQUANALTM home