Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > AQUANALTM Water Analysis > Learning Center > Compact Labs > Analytical Case for Building Restoration
AQUANALTM
Analytical Case for Building Restoration
Concept
AQUANAL Building Restoration
The analytical case for building restoration is designed for investigating the efflorescing of masonry, for aiding decontamination / redevelopment work and for analysing water and etravasted substances in soil in respect of constituents harmful to buildings.
The case contains tests for the determination of chloride, total hardness, magnesium, nitrate, sulfate and sulfide.
The operating manual gives advise how to prepare samples and shows how to calculate the results.
The evaluation of the results can either be done with Spectro 1 photometer or with Spectro 2 photometer - the photometers must be orded seperately!
Ordering Information
Product No.
Product Name
Add to Cart
37563
AQUANALTM-plus analytical case for building restoration
 :  
37560
Spectro 1 photometer for AQUANALTM-plus-reagent
 :  
37781
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue German
 :  
37782
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue English
 :  
37783
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue French
 :  
37784
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue Italian
 :  
37785
Spectro 2 photometer for AQUANALTM-plus-reagent, Menue Spanish
 :  
37441
refill pack chloride reagents
 :  
37456
refill pack total hardness reagents
 :  
37466
refill pack magnesium reagents
 :  
37449
refill pack nitrate reagents
 :  
37469
refill pack sulfate reagents
 :  
37470
refill pack sulfide reagents
 :  
back to top
Back to top
Compact Labs home
AQUANALTM home