 |
The nine ingredients for an aqueous living environment can be determined directly in lakes, rivers and ponds or at home in an aquarium using the AQUANALTM-plus Fishwater Lab contains six coloumetric and two titrimetric tests, the pH-value being determined by means of pH-paper. The evaluation is made using a colour card. More detailed information about the parameters and the effect of this kit on fish and plant life in inland waters can be found in the instructions for use. |
Parameters
Ammonium: is present in inland waters in a pH-dependant chemical equilibrium with ammonia. The test determines the total sum of ammonia and ammonium. Whilst ammonium is not dangerous for fish and is a fertiliser for plants, ammonia is a strong fish poison.
Carbonate Hardness: The fertility of a stretch of inland water can be ascertained with the help of carbonate hardness. This in fact dictates the quantity of fish present in the water. The acid-binding-capacity can, by association, also be determined using a multiplication factor.
Iron: Iron is an important factor; it is absolutely necessary for the production of haemoglobin but it can be dangerous for the entire fish population if it precipitates out in oxygen poor water. If this is the case, the gills become blocked, especially in young fish, and breathing is hindered.
Total Hardness: Determination of the total concentration of calcium and magnesium ions.
Nitrate: Nitrate can be washed into waterways from heavily fertilised fields. A high nitrate concentration leads to algae bloom and results in the decline of oxygen concentration.
Nitrite: In pure, healthy water, nitrite is never present. The presence of nitrite points to heavy pollution. Nitrite exists as an intermediate of natural decomposition and conversion processes of ammonium and nitrate. It indicates insufficient quality in the water necessary for fish.
Phosphate: On the one hand phosphate is a very important component of the skeleton and additionally for the energy exchange in plants and fish in water. However too much phosphate leads to over fertilisation and as a result to an oxygen deficiency.
pH-Value: The pH-value is influenced by most of the substances dissolved in water. A pH-value of 6.0 to 8.0 is normal for fish-sustaining waters. Values outside this range can result in diminished growth, skeletal deformities, skin and gill damage and at worst the death of the entire fish population.
Oxygen: The oxygen dissolved in water is essential for fish to breathe. Therefore the oxygen content should be as high as possible. The amount depends on, amongst other things, the temperature and bioorganisms in the water.
Measurement Limits of AQUANALTM-plus Fishwater Lab
| Parameter |
Measurement Limits |
Graduation of individual Tests |
Suitable for Sea Water |
| Ammonium |
≤0.05 - 10.0 mg/l |
≤0.05/ 0.2/ 1.0/ 3.0/ 10.0 |
no
|
| Carbonate Hardness |
≥1°CH |
1 drop = 1°CH |
yes
|
| Iron |
<0.05 - 1.0 mg/l |
<0.05/ 0.1/ 3.0/ 0.6/ 1.0 |
yes
|
| Total Hardness |
≥1°dH |
1 drop = 1°dH |
no
|
| Nitrate |
0 - 80 mg/l |
0/ 10/ 25/ 50/ 80 |
no
|
| Nitrite |
≤0.02 - 1.0 mg/l |
≤0.02/ 0.1/ 0.3/ 0.5/ 1.0 |
yes
|
| Phosphate |
0 - ≥6.0 mg/l |
0/ 0.5/ 1.2/ 3.0/ ≤6.0 |
yes
|
| pH-Value |
4.5 - 10.0 |
in 0.5 pH graduations |
yes
|
| Oxygen |
0 - 10 mg/l |
0/ 1/ 3/ 6/ 10 |
no
|
Ordering Information
| Product No. |
Product Name |
Add to Cart
|
| 37566 |
AQUANALTM-Fishwater Lab Case |
|
| 37550 |
AQUANALTM-Fishwater Lab Refill Pack complete |
|
|
Refill for single parameters of AQUANALTM-Fishwater Lab
|
| 37596 |
AQUANALTM-Fishwater Lab Carbonate Hardness single reagent |
|
| 37597 |
AQUANALTM-Fishwater Lab Iron single reagent |
|
| 37609 |
AQUANALTM-Fishwater Lab Oxygen single reagents |
|
| 37028 |
Special Indicator sticks, pH 4.0 - 10.0 |
|
| 37595 |
AQUANALTM-Ökotest Ammonium single reagents |
|
| 37598 |
AQUANALTM-Ökotest Total Hardness single reagent |
|
| 37599 |
AQUANALTM-Ökotest Nitrate single reagents |
|
| 37606 |
AQUANALTM-Ökotest Nitrite single reagent |
|
| 37607 |
AQUANALTM-Ökotest Phosphate single reagents |
|
| 37608 |
AQUANALTM-Ökotest pH single reagent |
|
| 37574 |
AQUANALTM-Ökotest Total Hardness reagent 250 ml |
|
| 37542 |
AQUANALTM-Fishwater Lab test tubes refill pack |
|
Back to top Compact Labs home AQUANALTM home
|