Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > AQUANALTM Water Analysis > Learning Center > Compact Labs > Pool Tester
AQUANALTM
Pool Tester
Concept
AQUANAL Pooltester
A high grade test kit with a big measuring range for determination of both free chlorine and pH-value in the practical hinged box which can be hung up.
The reading of the results is effected with the help of a color comparator.
The tablets are individually packed in aluminium foil. With the original pack (including the color comparator) 20 tests (each chlorine and pH-value) can be done, with refill pack (only including tablets) 30 tests can be done.
Ordering Information
Product No.
Product Name
Add to Cart
37546
AQUANALTM-Pooltester testkit (20 determinations each)
37547
AQUANALTM-Pooltester Refill Pack (30 determinations)
Back to top
Compact Labs home
AQUANALTM home