| Ordering Information |
| Catalog No. |
Description |
Add to Cart |
|
Accessories
|
| 37438 |
Test Tubes, Syringues, Spatula |
|
|
Acid Capacity, suitable for sea water
|
| |
Range:
|
0.1 = 1 mmol/l |
|
| 37415 |
Titrations Set suitable for 200 determinations |
|
| 37455 |
Refill Pack for 37415 |
|
| 37798 |
Check Solution 10 mmol/l for 37415 |
|
|
Aluminium (Al)
|
|
Range:
|
0.02 - 0.2 mg/l visual 0.02 - 0.3 mg/l photometric
|
|
| 37425 |
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations |
|
| 37465 |
Refill Pack for 37425 |
|
| 37505 |
Check Solution for 37425 |
|
|
Ammonium (NH4)
|
|
Range: |
0.2 - 8.0 mg/l visual 0.1 - 3.0 mg/l photometric
|
|
| 37400 |
Test Kit with Colour Comparator, Check solution: suitable for 250 determinations |
|
| 37440 |
Refill Pack for 37400 |
|
| 37480 |
Check Solution for 37400 |
|
|
Basic Capacity
|
|
Range: |
0.1 ml = 1 mmol
|
|
| 37414 |
Titrations Set suitable for 200 determinations |
|
| 37454 |
Refill Pack for 37414 |
|
| 37480 |
Check Solution 10 mmol/l for 37414 |
|
|
Cadmium (Cd)
|
|
Range: |
0.02 - 1.2 mg/l visual 0.02 - 1.0 mg/l photometric
|
|
| 37432 |
Test Kit with Colour Comparator, Check solution: suitable for 120 determinations |
|
| 37472 |
Refill Pack for 37432 |
|
| 37480 |
Check Solution for 37432 |
|
|
Carbonate Hardness, suitable for sea water
|
|
Range: |
1 drop = 1°CH
|
|
| 37437 |
Titrations Set suitable for 60 determinations approx. 15°CH |
|
| 37544 |
Refill Pack for 37437 |
|
| 37791 |
Check Solution 5°CH for 37437 |
|
| 37792 |
Check Solution 10°CH for 37437 |
|
|
Chlorine (Cl2), suitable for sea water
|
|
Range: |
0.01 - 0.30 mg/l visual 0.05 - 1.0 mg/l photometric
|
|
| 37419 |
Test Kit with Colour Comparator, Check solution: suitable for 300 determinations |
|
| 37459 |
Refill Pack for 37419 |
|
| 37499 |
Check Solution for 37419 |
|
|
Chlorid (Cl)
|
|
Range: |
5 - 300 mg/l visual 2 - 100 mg/l photometric
|
|
| 37401 |
Test Kit with Colour Comparator, Check solution: suitable for 300 determinations |
|
| 37441 |
Refill Pack for 37401 |
|
| 37481 |
Check Solution for 37401 |
|
| Chromium (Cr), suitable for sea water |
| |
Range:
|
0.005 - 0.1 mg/l visual 0.02 - 0.25 mg/l photometric
|
|
| 37402 |
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations |
|
| 37442 |
Refill Pack for 37402 |
|
| 37499 |
Check Solution for 37402 |
|
|
Cyanide (CN)
|
| |
Range:
|
0.03 - 0.7 mg/l visual 0.01 - 0.5 mg/l photometric
|
|
| 37403 |
Test Kit with Colour Comparator suitable for 200 determinations |
|
| 37443 |
Refill Pack for 37403 |
|
| Cyanide/Thiocyanate (CN/SCN), suitable for sea water |
| |
Range:
|
0.03 - 0.7 mg/l visual 0.01 - 0.5 mg/l photometric
|
|
| 37420 |
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations |
|
| 37460 |
Refill Pack for 37420 |
|
| 37500 |
Check Solution for 37420 |
|
|
Iron (Fe), suitable for sea water
|
| |
Range:
|
0.02 - 0.2 mg/l visual 0.02 - 0.2 mg/l photometric
|
|
| 37421 |
Test Kit with Colour Comparator, Check solution: suitable for 100 determinations |
|
| 37461 |
Refill Pack for 37421 |
|
| 37501 |
Check Solution for 37421 |
|
|
Iron (Fe), suitable for sea water
|
| |
Range:
|
0.2 - 15 mg/l visual 0.1 - 15 mg/l photometric
|
|
| 37404 |
Test Kit with Colour Comparator, Check solution: suitable for 500 determinations |
|
| 37444 |
Refill Pack for 37404 |
|
| 37484 |
Check Solution for 37404 |
|
|
Total Hardness
|
| |
Range:
|
1 drop = 1°d (0.178 mmol/l Ca)
|
|
| 37416 |
Titration Sets suitable for 500 determinations approx. 15°d |
|
| 37456 |
Refill Pack for 37416 |
|
| 37793 |
Check Solution 5°dH for 37416 |
|
| 37794 |
Check Solution 10°dH for 37416 |
|
| 37574 |
One-component reagent 250 ml |
|
|
Gold (Au), suitable for sea water
|
| |
Range:
|
0.1 - 3.0 mg/l
|
|
| 37537 |
Test Kit for photometric Analysis suitable for 140 determinations |
|
| Hydrazinium (N2H4) |
| |
Range:
|
0.02 - 1.05 mg/l visual 0.02 - 3.0 mg/l photometric
|
|
| 37433 |
Test Kit with Colour Comparator suitable for 130 determinations |
|
| 37473 |
Refill Pack for 37433 |
|
|
Copper (Cu), suitable for sea water
|
| |
Range:
|
0.05 - 0.5 and 0.4 - 4.5 mg/l visual 0.1 - 5.0 mg/l photometric
|
|
| 37424 |
Test Kit with Colour Comparator, Check solution: suitable for 75 determinations |
|
| 37464 |
Refill Pack for 37424 |
|
| 37504 |
Check Solution for 37424 |
|
|
Magnesium (Mg), suitable for sea water
|
| |
Range:
|
100 - 1500 mg/l visual 100 - 1500 mg/l photometric
|
|
| 37426 |
Test Kit with Colour Comparator, Check solution: suitable for 60 determinations |
|
| 37466 |
Refill Pack for 37426 |
|
| 37506 |
Check Solution for 37426 |
|
|
Manganese (Mn)
|
| |
Range:
|
0.03 - 0.5 mg/l visual 0.05 - 2.0 mg/l photometric
|
|
| 37406 |
Test Kit with Colour Comparator, Check solution: suitable for 110 determinations |
|
| 37446 |
Refill Pack for 37406 |
|
| 37486 |
Check Solution for 37406 |
|
|
Nickel (Ni)
|
| |
Range:
|
0.02 - 0.5 mg/l visual 0.01 - 1.5 mg/l photometric
|
|
| 37407 |
Test Kit with Colour Comparator, Check solution: suitable for 125 determinations |
|
| 37447 |
Refill Pack for 37407 |
|
| 37487 |
Check Solution for 37407 |
|
| Nitrate (NO3) |
| |
Range:
|
5 - 140 mg/l visual 5 - 140 mg/l photometric
|
|
| 37408 |
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations |
|
| 37448 |
Refill Pack for 37408 |
|
| 37488 |
Check Solution for 37408 |
|
|
Nitrate (NO3)
|
| |
Range:
|
1 - 50 mg/l visual 0.5 - 50 mg/l photometric
|
|
| 37409 |
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations |
|
| 37449 |
Refill Pack for 37409 |
|
| 37489 |
Check Solution for 37409 |
|
|
Nitrite (NO2), suitable for sea water
|
| |
Range:
|
0.005 - 0.1 mg/l visual 0.02 - 1.0 mg/l photometric
|
|
| 37410 |
Test Kit with Colour Comparator suitable for 110 determinations |
|
| 37450 |
Refill Pack for 37410 |
|
|
Oxygen (O2)
|
| |
Range:
|
1 - 12 mg/l visual 0.5 - 12 mg/l photometric
|
|
| 37428 |
Test Kit with Colour Comparator suitable for 30 determinations |
|
| 37468 |
Refill Pack for 37428 |
|
|
Oxygen Scavenger
|
| |
Range:
|
0.2 - 3.0 mg/l
|
|
| 37418 |
Test Kit with Colour Comparator suitable for 100 determinations |
|
| 37458 |
Refill Pack for 37418 |
|
|
Peroxide (H2O2), suitable for sea water
|
| |
Range:
|
1 - 30 mg/l visual 1 - 100 mg/l photometric
|
|
| 37434 |
Test Kit with Colour Comparator suitable for 180 determinations |
|
| 37474 |
Refill Pack for 37434 |
|
|
Phenol (C6H5OH), suitable for sea water
|
| |
Range:
|
0.1 - 3.0 mg/l visual 0.1 - 3.0 mg/l photometric
|
|
| 37427 |
Test Kit with Colour Comparator suitable for 300 determinations |
|
| 37467 |
Refill Pack for 37427 |
|
|
Phosphate (PO4), suitable for sea water
|
| |
Range:
|
3 - 120 mg/l visual 3 - 120 mg/l photometric
|
|
| 37423 |
Test Kit with Colour Comparator, Check solution: suitable for 210 determinations |
|
| 37463 |
Refill Pack for 37423 |
|
| 37503 |
Check Solution for 37423 |
|
|
Phosphate (PO4), suitable for sea water
|
| |
Range:
|
0.02 - 0.4 mg/l visual 0.1 - 2.0 mg/l photometric
|
|
| 37411 |
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations |
|
| 37451 |
Refill Pack for 37411 |
|
| 37491 |
Check Solution for 37411 |
|
|
Residual Hardness
|
| |
Range:
|
1 drop = 0.01°d (0.00178 mmol/l Ca)
|
|
| 37417 |
Titrations Set suitable for 340 determinations |
|
| 37457 |
Refill Pack for 37417 |
|
| 37795 |
Check Solution 0.05°dH for 37417 |
|
| 37796 |
Check Solution 0.10°dH for 37417 |
|
|
Silver (Ag)
|
| |
Range: |
0.1 - 1.0 mg/l
|
|
| 37538 |
Test Kit for Photometric Analysis suitable for 300 determinations |
|
|
Silicium (Si) |
| |
Range:
|
0.01 - 0.25 mg/l visual 0.2 - 0.25 mg/l photometric
|
|
| 37412 |
Test Kit with Colour Comparator, Check solution: suitable for 150 determinations |
|
| 37452 |
Refill Pack for 37412 |
|
| 37492 |
Check Solution for 37412 |
|
|
|
| |
Range:
|
50 - 330 mg/l visual 40 - 500 mg/l photometric
|
|
| 37429 |
Test Kit with Colour Comparator, Check solution: suitable for 100 determinations |
|
| 37469 |
Refill Pack for 37429 |
|
| 37509 |
Check Solution for 37429 |
|
|
Sulfide (S2-)
|
| |
Range:
|
0.03 - 0.3 mg/l visual 0.05 - 1.0 mg/l photometric
|
|
| 37430 |
Test Kit with Colour Comparator suitable for 100 determinations |
|
| 37470 |
Refill Pack for 37430 |
|
|
Sulfite (SO3)
|
| |
Range:
|
0.1 ml = 10 mg/l
|
|
| 37435 |
Titrations Set suitable for 150 determinations |
|
| 37475 |
Refill Pack for 37435 |
|
|
Tin (Sn)
|
|
Range:
|
0.1 - 1.2 mg/l visual 0.1 - 1.2 mg/l photometric
|
|
| 37436 |
Test Kit with Colour Comparator suitable for 120 determinations |
|
| 37476 |
Refill Pack for 37436 |
|
|
Water for Dilution
|
| 37439 |
Bottle with 90 ml |
|
|
Zinc (Zn)
|
| |
Range:
|
0.1 - 5.0 mg/l visual
|
|
| 37413 |
Test Kit with Colour Comparator, Check solution: suitable for 120 determinations |
|
| 37453 |
Refill Pack for 37413 |
|
| 37493 |
Check Solution for 37413 |
|
|
Zinc (Zn)
|
| |
Range:
|
0.02 - 0.2 mg/l
|
|
| 37539 |
Test Kit for Photometric Analysis suitable for 120 determinations |
|