Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > AQUANALTM Water Analysis > Test Kits
AQUANALTM
Test Kits
Test kits for approximately 40 substances
•
•
•
•
•
•
titrimetric or coloimetric analysis
each test kit allows between 100 and
500 determinations
refill packs available
quick and easy handling
standard solution included to check the function
of the reagents
no laboratory equipment required, everything
necessary to do the tests is included
results can be taken...
visually:
- with colour chart
- coloured dots in sensible graduation
- exact enough to state: below or above limit
- most economical solution
photometrically:
- independent of light influences
- result is given in extinction values or mg/l
- reliable values, reproducible by various persons
  (not influenced by varying judgements about colours)
Test kit for approximately 40 substances
Ordering Information
Catalog No.
Description
Add to Cart
Accessories
37438
Test Tubes, Syringues, Spatula
Acid Capacity, suitable for sea water
Range:
0.1 = 1 mmol/l
37415
Titrations Set suitable for 200 determinations
37455
Refill Pack for 37415
37798
Check Solution 10 mmol/l for 37415
Aluminium (Al)
Range:
0.02 - 0.2 mg/l visual
0.02 - 0.3 mg/l photometric
37425
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations
37465
Refill Pack for 37425
37505
Check Solution for 37425
Ammonium (NH4)
Range:
0.2 - 8.0 mg/l visual
0.1 - 3.0 mg/l photometric
37400
Test Kit with Colour Comparator, Check solution: suitable for 250 determinations
37440
Refill Pack for 37400
37480
Check Solution for 37400
Basic Capacity
Range:
0.1 ml = 1 mmol
37414
Titrations Set suitable for 200 determinations
37454
Refill Pack for 37414
37480
Check Solution 10 mmol/l for 37414
Cadmium (Cd)
Range:
0.02 - 1.2 mg/l visual
0.02 - 1.0 mg/l photometric
37432
Test Kit with Colour Comparator, Check solution: suitable for 120 determinations
37472
Refill Pack for 37432
37480
Check Solution for 37432
Carbonate Hardness, suitable for sea water
Range:
1 drop = 1°CH
37437
Titrations Set suitable for 60 determinations approx. 15°CH
37544
Refill Pack for 37437
37791
Check Solution 5°CH for 37437
37792
Check Solution 10°CH for 37437
Chlorine (Cl2), suitable for sea water
Range:
0.01 - 0.30 mg/l visual
0.05 - 1.0 mg/l photometric
37419
Test Kit with Colour Comparator, Check solution: suitable for 300 determinations
37459
Refill Pack for 37419
37499
Check Solution for 37419
Chlorid (Cl)
Range:
5 - 300 mg/l visual
2 - 100 mg/l photometric
37401
Test Kit with Colour Comparator, Check solution: suitable for 300 determinations
37441
Refill Pack for 37401
37481
Check Solution for 37401
Chromium (Cr), suitable for sea water
Range:
0.005 - 0.1 mg/l visual
0.02 - 0.25 mg/l photometric
37402
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations
37442
Refill Pack for 37402
37499
Check Solution for 37402
Cyanide (CN)
Range:
0.03 - 0.7 mg/l visual
0.01 - 0.5 mg/l photometric
37403
Test Kit with Colour Comparator suitable for 200 determinations
37443
Refill Pack for 37403
Cyanide/Thiocyanate (CN/SCN), suitable for sea water
Range:
0.03 - 0.7 mg/l visual
0.01 - 0.5 mg/l photometric
37420
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations
37460
Refill Pack for 37420
37500
Check Solution for 37420
Iron (Fe), suitable for sea water
Range:
0.02 - 0.2 mg/l visual
0.02 - 0.2 mg/l photometric
37421
Test Kit with Colour Comparator, Check solution: suitable for 100 determinations
37461
Refill Pack for 37421
37501
Check Solution for 37421
Iron (Fe), suitable for sea water
Range:
0.2 - 15 mg/l visual
0.1 - 15 mg/l photometric
37404
Test Kit with Colour Comparator, Check solution: suitable for 500 determinations
37444
Refill Pack for 37404
37484
Check Solution for 37404
Total Hardness
Range:
1 drop = 1°d (0.178 mmol/l Ca)
37416
Titration Sets suitable for 500 determinations approx. 15°d
37456
Refill Pack for 37416
37793
Check Solution 5°dH for 37416
37794
Check Solution 10°dH for 37416
37574
One-component reagent 250 ml
Gold (Au), suitable for sea water
Range:
0.1 - 3.0 mg/l
37537
Test Kit for photometric Analysis suitable for 140 determinations
Hydrazinium (N2H4)
Range:
0.02 - 1.05 mg/l visual
0.02 - 3.0 mg/l photometric
37433
Test Kit with Colour Comparator suitable for 130 determinations
37473
Refill Pack for 37433
Copper (Cu), suitable for sea water
Range:
0.05 - 0.5 and 0.4 - 4.5 mg/l visual
0.1 - 5.0 mg/l photometric
37424
Test Kit with Colour Comparator, Check solution: suitable for 75 determinations
37464
Refill Pack for 37424
37504
Check Solution for 37424
Magnesium (Mg), suitable for sea water
Range:
100 - 1500 mg/l visual
100 - 1500 mg/l photometric
37426
Test Kit with Colour Comparator, Check solution: suitable for 60 determinations
37466
Refill Pack for 37426
37506
Check Solution for 37426
Manganese (Mn)
Range:
0.03 - 0.5 mg/l visual
0.05 - 2.0 mg/l photometric
37406
Test Kit with Colour Comparator, Check solution: suitable for 110 determinations
37446
Refill Pack for 37406
37486
Check Solution for 37406
Nickel (Ni)
Range:
0.02 - 0.5 mg/l visual
0.01 - 1.5 mg/l photometric
37407
Test Kit with Colour Comparator, Check solution: suitable for 125 determinations
37447
Refill Pack for 37407
37487
Check Solution for 37407
Nitrate (NO3)
Range:
5 - 140 mg/l visual
5 - 140 mg/l photometric
37408
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations
37448
Refill Pack for 37408
37488
Check Solution for 37408
Nitrate (NO3)
Range:
1 - 50 mg/l visual
0.5 - 50 mg/l photometric
37409
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations
37449
Refill Pack for 37409
37489
Check Solution for 37409
Nitrite (NO2), suitable for sea water
Range:
0.005 - 0.1 mg/l visual
0.02 - 1.0 mg/l photometric
37410
Test Kit with Colour Comparator suitable for 110 determinations
37450
Refill Pack for 37410
Oxygen (O2)
Range:
1 - 12 mg/l visual
0.5 - 12 mg/l photometric
37428
Test Kit with Colour Comparator suitable for 30 determinations
37468
Refill Pack for 37428
Oxygen Scavenger
Range:
0.2 - 3.0 mg/l
37418
Test Kit with Colour Comparator suitable for 100 determinations
37458
Refill Pack for 37418
Peroxide (H2O2), suitable for sea water
Range:
1 - 30 mg/l visual
1 - 100 mg/l photometric
37434
Test Kit with Colour Comparator suitable for 180 determinations
37474
Refill Pack for 37434
Phenol (C6H5OH), suitable for sea water
Range:
0.1 - 3.0 mg/l visual
0.1 - 3.0 mg/l photometric
37427
Test Kit with Colour Comparator suitable for 300 determinations
37467
Refill Pack for 37427
Phosphate (PO4), suitable for sea water
Range:
3 - 120 mg/l visual
3 - 120 mg/l photometric
37423
Test Kit with Colour Comparator, Check solution: suitable for 210 determinations
37463
Refill Pack for 37423
37503
Check Solution for 37423
Phosphate (PO4), suitable for sea water
Range:
0.02 - 0.4 mg/l visual
0.1 - 2.0 mg/l photometric
37411
Test Kit with Colour Comparator, Check solution: suitable for 200 determinations
37451
Refill Pack for 37411
37491
Check Solution for 37411
Residual Hardness
Range:
1 drop = 0.01°d (0.00178 mmol/l Ca)
37417
Titrations Set suitable for 340 determinations
37457
Refill Pack for 37417
37795
Check Solution 0.05°dH for 37417
37796
Check Solution 0.10°dH for 37417
Silver (Ag)
Range:
0.1 - 1.0 mg/l
37538
Test Kit for Photometric Analysis suitable for 300 determinations
Silicium (Si)
Range:
0.01 - 0.25 mg/l visual
0.2 - 0.25 mg/l photometric
37412
Test Kit with Colour Comparator, Check solution: suitable for 150 determinations
37452
Refill Pack for 37412
37492
Check Solution for 37412
Sulfate (SO4)
Range:
50 - 330 mg/l visual
40 - 500 mg/l photometric
37429
Test Kit with Colour Comparator, Check solution: suitable for 100 determinations
37469
Refill Pack for 37429
37509
Check Solution for 37429
Sulfide (S2-)
Range:
0.03 - 0.3 mg/l visual
0.05 - 1.0 mg/l photometric
37430
Test Kit with Colour Comparator suitable for 100 determinations
37470
Refill Pack for 37430
Sulfite (SO3)
Range:
0.1 ml = 10 mg/l
37435
Titrations Set suitable for 150 determinations
37475
Refill Pack for 37435
Tin (Sn)
Range:
0.1 - 1.2 mg/l visual
0.1 - 1.2 mg/l photometric
37436
Test Kit with Colour Comparator suitable for 120 determinations
37476
Refill Pack for 37436
Water for Dilution
37439
Bottle with 90 ml
Zinc (Zn)
Range:
0.1 - 5.0 mg/l visual
37413
Test Kit with Colour Comparator, Check solution: suitable for 120 determinations
37453
Refill Pack for 37413
37493
Check Solution for 37413
Zinc (Zn)
Range:
0.02 - 0.2 mg/l
37539
Test Kit for Photometric Analysis suitable for 120 determinations
Back to top
AQUANALTM home