Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > AQUANALTM Water Analysis > Test Sticks
AQUANALTM
Test Sticks
QUANTOFIX Test Sticks
Concept
Quantofix® test sticks serve for semi-quantitative determination of ions and molecules. The benefits of using Quantofix® test sticks tocarry out a determination are evident: it is quick -dip the test stick and read the result- and easy -no further equipment needed-. A number of determinations require the use of additional reagents which are enclosed with the test sticks in a single kit. Instructions for use are enclosed as well. The test sticks themselves are about 0.2 mm thick and made of plastic; each pack allows 100 tests to be carried out. Our product range includes the following tests:
Test
Measuring Range
Graduation of
Color Scale in mg/L
Color Reaction
Catalog No.
Aluminium
0-500 mg/l Al3+
0-5-20-200-500
pink --> red
37207
Ammonium
0-400 mg/l NH4+
0-10-25-50-100-200-400
yellow --> orange
37212
Arsenic
0 - 0.5 mg/l As3+/5+
0 - 0.01 - 0.025 - 0.05 - 0.1 - 0.5
white --> yellowish-brown
37927
Ascorbic Acid
0-2000 mg/l Vitamin C
0-5-100-200-300-500-1000-2000
yellow --> bluish green
37203
Chlorine
0-100 mg/l Cl2
0-1-3-10-30-100
white --> blue
37208
Chromate
0-100 mg/l CrO42-
0-3-10-30-100
pink --> violet
37209
Cobalt
0-1000 mg/l Co2+
0-10-25-50-100-250-500-1000
white --> greenish blue
37199
Copper
0-300 mg/L Cu2+
0-10-30-100-300
white --> red violet
37198
Fluoride
0-100 mg/l F-
0–2–5–10–20–50–100
red --> colorless
37211
Nickel
0-1000 mg/l Ni2+
0-10-25-50-100-250-500-1000
white --> red
37197
Nitrate (Nitrite)
0-500 mg/l NO3-
Nitrate: 0-10-25-50-100-250-500
white --> pink
37204
0-80 mg/l NO2-
Nitrite: 0-1-5-10-20-40-80
Nitrite
0-50 mg/l NO2-
0-1-5-10-25-50
white --> pink
37205
Peroxide
0-100 mg/l H2O2
0-1-3-10-30-100
white --> blue
37206
Phosphate
0-100 mg/l PO43-
0-3-10-25-50-100
white --> greenish blue
37210
Potassium
0-1500 mg/l K+
0-200-400-700-1000-1500
yellow --> orange
37202
Silver (Ag-Fix)
0-10 g/l Ag+
0-0.5-1-2-3-7-10
ochreous --> brown
37196
pH value 4-8
pH value: 4-5-6-7-8
yellow --> blue
Sulfate
0-1600 mg/l SO42-
0-200-400-800-1200-1600
red --> yelow
37213
Sulfite
0-1000 mg/l SO32-
0-10-25-50-100-250-500-1000
white --> pink
37201
Tin
0-500 mg/l Sn2+
0-10-25-50-100-250-500
white --> blue
37195
Water Hardness - Aquadur®
3-21°dH
0 – 4 – 7 – 14 – 21 °d
turquoise --> pink
37194
button-labinfos
Back to top
AQUANALTM home