|
| |
The Thermoblock is used to heat the tubes during the decomposition. It works at a fixed temperature, either 100°C or 148°C. With the help of a timer function fixed reaction times can be given. A beeper will remind you of the end of reaction time, the Thermoblock will be automatically switched off.
|
| Technical data |
| Temperature |
100°C and 148°C fixed, can be selected via a membrane keyboard |
| Tolerance |
± 1°C |
| Heating |
300 W, electronically regulated, with protection against overheating |
| Heating block |
Eight wells for tubes of 16 mm diameter, 1 well in the centre for tubes of 22 mm diameter |
| Time input |
30, 60, 120 minutes or continuous operation at the selected temperature, automatically switched off at the end of the selected reaction time, acoustic signal |
| Casing |
Round metal block with epoxide-coated metal casing |
| Dimensions |
D x W x H 214 x 1343 x 110 mm, supplied in a cardboard box |
| Weight |
2.15 kg |
| Mains connection |
230 V, 50 Hz (350 W) or 110 V, 60 Hz |
| Plug |
Euroformat |
| Safety |
CE marking, transparent covering hood |
| Guarantee |
1 year from date of sale |
| Product # |
Product Name |
Add to Cart |
| 37571 |
AQUANAL-plus Thermoblock |