Sigma-Aldrich
Analytical / Chromatography > Analytical Reagents > AQUANALTM Water Analysis > Thermoblock
AQUANALTM
Thermoblock
AQUANAL Thermoblock heating unit
The Thermoblock is used to heat the tubes during the decomposition. It works at a fixed temperature, either 100°C or 148°C. With the help of a timer function fixed reaction times can be given. A beeper will remind you of the end of reaction time, the Thermoblock will be automatically switched off.
Technical data
Temperature 100°C and 148°C fixed, can be selected via a membrane keyboard
Tolerance ± 1°C
Heating 300 W, electronically regulated, with protection against overheating
Heating block Eight wells for tubes of 16 mm diameter, 1 well in the centre for tubes of 22 mm diameter
Time input 30, 60, 120 minutes or continuous operation at the selected temperature, automatically switched off at the end of the selected reaction time, acoustic signal
Casing Round metal block with epoxide-coated metal casing
Dimensions D x W x H 214 x 1343 x 110 mm, supplied in a cardboard box
Weight 2.15 kg
Mains connection 230 V, 50 Hz (350 W) or 110 V, 60 Hz
Plug Euroformat
Safety CE marking, transparent covering hood
Guarantee 1 year from date of sale
Product #  Product Name Add to Cart
37571 AQUANAL-plus Thermoblock