AQUANAL®
Vario H Powder Packs
Introduction
Use
Chlorine, Free and Total
Product List
Introduction
Vario H Powder Packs are designed to be a like for like chemistry replacing Hach® powder pillows.
The use of dry powdered reagents minimises leakage and deterioration problems. Powders are packed in individual, pre-measured foil packs. Each pack contains enough reagent for one test ensuring identical test results compared to Hach® Powder pillows.
Variop H Powder Packs
Use of Vario H Powder Packs
Use of Vario H Powder Packs
Back to top
Chlorine, free and total
  • Free Chlorine
    Free Chlorine (Hypochlorous Acid and Hypochlorite Ion) oxidizes DPD. The reactions is pH dependent. DPD and a buffer system (pH 6.3-6.5) cause a magenta colour in the presence of Free Chlorine. Both, our Vario H Powder and Hach® Powder handle high levels of hardness without precipitation.
  • Total Chlorine
    Potassium Iodide is used to determine combined chlorine forms. Chloramines oxidize the iodide to iodine; the liberated iodine reacts with DPD. Both, Sigma-Aldrich Vario H Powder and Hach® Powder contain DPD, Potassium Iodide and a buffer system.
Back to top
Product List
Product #
Range
Product Name
HACH® Prod.No.
Add to Cart
Aluminium
70436
0-0.22 mg/l Al AQUANAL®-professional Vario H Aluminium Set 26037-00
Chlorine, free
70112
0.01-2 mg/l Cl2 AQUANAL®-professional Vario H Chlorine Free (DPD) for 10 ml sample 21055-69
70113
0-5 mg/l AQUANAL®-professional Vario H Chlorine Free (DPD), for 25 ml sample 14070-99
Chlorine, total
70200
0.01-2 mg/l Cl2 AQUANAL®-professional Vario H Chlorine Total (DPD) for 10 ml sample 21056-69
70202
0-5 mg/l Cl2 AQUANAL®-professional Vario H Chlorine Total (DPD), for 25 ml sample 14064-99
Copper
70203
0-5 mg/l Cu AQUANAL®-professional Vario H Copper I 21058-69
Iron, total
70204
0-1.8 mg/l Fetot AQUANAL®-professional Vario H Iron (TPTZ) for 10 ml sample 26087-99
70205
0-3 mg/l Fetot AQUANAL®-professional Vario H Iron (Phenantroline), for 10 ml sample 21057-69
Manganese LR
70441
0-0.7 mg/l Mn AQUANAL®-professional Vario H Manganese Set, low range 26517-00
Molybdate HR
70209
0-35 mg/l Mo AQUANAL®-professional Vario H Molybdate Set, high range for 10 ml sample 26041-00
70426
  AQUANAL®-professional Vario H Molybdate Set, high range for
25 ml sample
22434-00
Nitrate
70431
0-30 mg/l NO3-N AQUANAL®-professional Vario H Nitrate Set, high range 26053-45
Nitrite LR
70219
0-0.3 mg/l NO2-N AQUANAL®-professional Vario H Nitrite for 25 ml sample 14065-99
70218
0-0.3 mg/l NO2-N AQUANAL®-professional Vario H Nitrite for 10 ml sample 21071-69
Nitrogen, Ammonia LR
70434
0-2.5 mg/l NH3-N AQUANAL®-professional Vario H Ammonia Set, low range 26045-45
Nitrogen, Ammonia HR
70423
0-50 mg/l NH3-N AQUANAL®-professional Vario H Ammonia Set, high range 26069-45
70222
  AQUANAL®-professional Vario H Ammonia Cyanurate, refill pack for 70434 and 70423 23954-66
70223
  AQUANAL®-professional Vario H Ammonia Salicylate, refill pack for 70434 and 70423 23952-66
Nitrogen, Total HR
70412
10-150 mg/l N AQUANAL®-professional Vario H Nitrogen Set, high range 27141-00
o-Phosphate
70229
0-2.5 mg/l PO4 AQUANAL®-professional Vario H Phosphate for 10 ml sample 21060-69
Silica LR
70417
0-1.6 mg/l SiO2 AQUANAL®-professional Vario H Silica Set, low range 24593-00
Silica HR
70232
0-100 mg/l SiO2 AQUANAL®-professional Vario H Silica Set, high range 24596-00
Sulfate
70244
0-70 mg/l SO4 AQUANAL®-professional Vario H Sulfate for 25 ml sample 12065-99
70421
  AQUANAL®-professional Vario H Sulfate for 10 ml sample 21067-69
Hach® is a registered trademark of the Hach Company
button-labinfos
Back to top
AQUANAL® home