AQUANAL®
Waste Water Analysis
For your convenience we offer ready-to-use solutions according to the "European Standard Methods for Water Analysis" (waste water, sewage water).
Ready-to-use solutions for waste water analysis
Ordering Information
Product #
Product Name
Package Size
Add to Cart
32028
Fluoride buffer solution 1 l
34621
Mercury sulfate solution (20 g/l) 1 l
34632
Mercury sulfate solution (80 g/l) 1 l and 2.5 l
34624
Mercury sulfate solution (200 g/l) 1 l
33584
Phenolindex Buffer Solution pH 10 1 l
33585
Phenolindex Buffer Solution pH 4 1 l
34629
Silver sulfate solution (10 g/l) 1 l and 2.5 l
31066
Silver sulfate solution (60 g/l) 1 l
31067
Silver sulfate solution (80 g/l) 1 l and 2.5 l
34244
Titanylsulfate solution 1 l