Gas Chromatography
GC Derivatizing Agents
Why is chemical derivatization needed?
Gas chromatography, a technique for separation of volatile compounds which are thermally stable, is unfortunately not always easily to apply for compounds of biomedical and environmental interest, particularly for those of high molecular weight or containing polar functional groups. These groups are difficult to analyze by GC either because they are not sufficiently volatile, tail badly, are too strongly attracted to the stationary phase, thermally unstable or even decomposed.
Chemical derivatization prior to analysis is generally done to:
  • increase the volatility and decrease the polarity of compounds;
  • reduce thermal degradation of samples by increasing their thermal stability;
  • increase detector response by incorporating functional groups which lead to higher detector signals, e.g. CF3 groups for electron capture detectors;
  • improve separation and reduce tailing
  • enlarge substrate spectrum
What kind of Derivatizing Reagents exist ?
Common derivatization methods can be classified into 4 groups depending on the type of reaction applied:
To view the list of derivatizing agents for all methods, please click on the method above.
Due to our broad experience in production and analysis of such derivatizing reagents and especially of silylating agents we can assure that you only get reliable, clearly specified and customer-focused products.
Silylating Reagents
Catalog No.
Product
CAS No.
Description
05940
Allyldimethylchlorosilane
4028-23-3
purum; ≥97.0% (GC)
09324
3-Aminopropyltriethoxysilane
919-30-2
purum; ~99% (GC)
09326
3-Aminopropyltrimethoxysilane
13822-56-5
purum; ≥97.0% (NT)
14660
1,3-Bis(chloromethyl)tetramethyl-disilazane
14579-91-0
purum; ~97% (GC)
14700
Bis(diethylamino)dimethylsilane
4669-59-4
purum; ~97% (GC)
14755
Bis(dimethylamino)dimethylsilane
3768-58-9
purum; ~97% (GC)
15242
N,O-Bis(trimethylsilyl)acetamide, 10x1ml
10416-59-8
purum; ~97% (GC)
15269
N,O-Bis(trimethylsilyl)acetamide, for GC
10416-59-8
purum p.a. for GC; ≥97.0% (GC)
15256
N,O-Bis(trimethylsilyl)acetamide with 5% TMCS, for GC
for GC
15236
N,O-Bis(trimethylsilyl)carbamate
35342-88-2
purum; ≥98.0% (T)
15247
N,N-Bis(trimethylsilyl)formamide
15500-60-4
pract.; ~95% (GC)
15235
N,N-Bis(trimethylsilyl)methylamine
920-68-3
purum; ≥97.0% (GC)
15249
Bis(trimethylsilyl)sulfate
18306-29-1
purum; ≥98.0% (T)
15198
N,O-Bis(trimethylsilyl)trifluoroacetamide, for GC, 10x1ml
25561-30-2
puriss. p.a. for GC; ≥99.0% (GC)
15222
N,O-Bis(trimethylsilyl)trifluoroacetamide, for GC
25561-30-2
puriss. p.a. for GC; ≥99.0% (GC)
15243
N,O-Bis(trimethylsilyl)trifluoroacetamide
25561-30-2
purum; ≥98.0% (GC)
15238
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% TMCS
25561-30-2
for GC
15239
N,O-Bis(trimethylsilyl)trifluoroacetamide with 1% TMCS 10x1ml
25561-30-2
for GC
15209
N,O-Bis(trimethylsilyl)trifluoroacetamide with 10% TMCS
95596-72-8
for GC
15248
N,N'-Bis(trimethylsilyl)urea
18297-63-7
purum; ≥98.0% (N)
17615
(Bromomethyl)dimethylchlorosilane
16532-02-8
purum; ~98% (GC)
19903
Butyldimethylchlorosilane
1000-50-6
purum; ≥97.0% (GC)
19904
tert-Butyldimethylchlorosilane Solution, ~1 M in THF
18162-48-6
purum; ~1 M in tetrahydrofuran
19920
1-(tert-Butyldimethylsilyl)imidazole
54925-64-3
purum; ~97% (GC)
19915
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide
77377-52-7
purum; ≥97.0% (GC)
19918
N-tert-Butyldime.silyl-N-me.trifluoroacetamide + 1% TBDMSCl
77377-52-7
purum
19938
tert-Butyldiphenylchlorosilane
58479-61-1
purum; ≥97.0% (GC)
25117
(Chloromethyl)dimethylchlorosilane
1719-57-9
purum; ~97% (GC)
26230
(3-Chloropropyl)dimethylchlorosilane
10605-40-0
pract.; ≥95% (GC)
26250
(3-Chloropropyl)trimethoxysilane
2530-87-2
purum; ~97% (GC)
28470
(2-Cyanoethyl)trichlorosilane
1071-22-3
purum; ≥98.0% (GC)
28560
3-Cyanopropyltrichlorosilane
1071-27-8
purum; ≥98.0% (GC)
28558
3-Cyanopropyltriethoxysilane
1067-47-6
purum; ≥96% (GC)
32575
N,N-Diethyltrimethylsilylamine
996-50-9
purum; ~97% (GC)
15226
(+)-Diisopropyl O,O'-bis(trimethylsilyl)-L-tartrate
130678-42-1
ChiraSelect; ~99% (GC, sum of enantiomers)
15223
(-)-Diisopropyl O,O'-bis(trimethylsilyl)-D-tartrate
197013-45-9
ChiraSelect; ~99% (GC, sum of enantiomers)
39900
Dimethylchlorosilane
1066-35-9
BioChemika purum; ~96% (GC)
40140
Dimethyldichlorosilane
75-78-5
BioChemika puriss.; ≥99,5% (GC)
40925
Dimethyl(3,3,4,4,5,5,6,6,6-nonafluorohe-xyl)chlorosil.(2 ml)
119386-82-2
purum; ≥97.0% (GC)
41663
1,3-Dimethyl-1,1,3,3-tetraphenyldi-silazane
7453-26-1
purum; ≥98.0% (NT)
41720
N,N-Dimethyltrimethylsilylamine
2083-91-2
purum; ~97% (GC)
43300
Diphenylsilanediol
947-42-2
purum; ≥97.0% (CH)
43340
1,3-Diphenyl-1,1,3,3-tetramethyl disila-zane
3449-26-1
purum; ≥97.0% (GC)
43930
1,3-Divinyl-1,1,3,3-tetramethyldisilazane
7691-02-3
purum; ~97% (GC)
44230
Dodecyltrichlorosilane
4484-72-4
purum; ~97% (GC)
44235
Dodecyltrichlorosilane
4484-72-4
puriss.; ≥99.0% (GC)
44237
Dodecyltriethoxysilane
18536-91-9
pract.; ≥95% (GC)
92756
Ethyl trimethylsilylacetate
4071-88-9
purum; ~97% (GC)
50040
(3-Glycidyloxypropyl)trimethoxysilane
2530-83-8
purum; ≥97.0% (GC)
52614
2,2,4,4,6,6-Hexamethylcyclotrisilazane
1009-93-4
purum; ≥97.0% (GC)
52619
Hexamethyldisilazane
999-97-3
puriss.; ≥99.0% (GC)
52630
Hexamethyldisiloxane
107-46-0
puriss.; ~99% (GC)
52643
Hexamethyldisilthiane
3385-94-2
purum
58810
(3-Isocyanatopropyl)triethoxysilane
24801-88-5
pract.; ≥95% (GC)
63797
(3-Mercaptopropyl)triethoxysilane
14814-09-6
techn.; ≥80% (GC)
63800
(3-Mercaptopropyl)trimethoxysilane
4420-74-0
purum; ≥97.0% (GC)
68210
Methyl-octyldichlorosilane
14799-93-0
purum; ≥98.0% (GC)
92758
(Methylthio)trimethylsilane
3908-55-2
purum; ≥97.0% (GC)
69450
Methyltrichlorosilane
75-79-6
puriss.; ≥99.0% (AT)
69480
N-Methyl-N-trimethylsilylacetamide
7449-74-3
purum; ≥97.0% (GC)
69484
N-Methyl-N-trimethylsilylheptafluorobutyramide
53296-64-3
pract.; ≥90% (GC)
69479
N-Methyl-N-trimethylsilyltrifluoroacet-amide, for GC
24589-78-4
puriss. for GC; ~99% (GC)
69482
N-Methyl-N-trimethylsilyltrifluoroacetamide
24589-78-4
purum; ≥97.0% (GC)
74762
Octadecyltrichlorosilane
112-04-9
pract.; ~90% (GC)
74891
Octaphenylcyclotetrasiloxane
546-56-5
purum; ≥98.0% (CH)
75085
Octyltrichlorosilane
5283-66-9
purum; ≥98.0% (GC)
76750
(Pentafluorophenyl)dimethylchlorosilane
20082-71-7
purum; ≥97.0% (GC)
85431
Silylating mixture Fluka I according to Sweeley for GC   for GC; ready to use solution in pyridine (10 vol parts)
85434
Silylating mixture Fluka I acc. to Sweeley (10x1 ml)   for GC; ready to use solution in pyridine (10 vol parts); ampoule
85432
Silylating mixture Fluka II acc. to Horning for GC   for GC
85435
Silylating mixture Fluka II acc. to Horning (10x1 ml)   for GC; ampoule
85433
Silylating mixture Fluka III for GC   for GC
85436
Silylating mixture Fluka III for GC(10x1 ml)   for GC; ampoule
87830
1,1,3,3-Tetramethyldisilazane
15933-59-2
purum; ≥97.0% (GC)
90383
Triethylchlorosilane
994-30-9
purum; ≥97.0% (GC)
90385
Triethylchlorosilane Solution,~1 M in THF
994-30-9
purum; ~1 M in tetrahydrofuran
91743
Triethylsilyl trifluoromethanesulfonate
79271-56-0
purum; ≥98.0% (T)
92090
Triisopropylchlorosilane
13154-24-0
purum; ~97% (GC)
91746
Triisopropylsilyl trifluoromethanesulfonate
80522-42-5
purum; ≥98.0% (T)
92337
Trimethylbromosilane, stab.
2857-97-8
purum; ≥97.0% (AT); stab. with silver wool
92360
Trimethylchlorosilane
75-77-4
puriss.; ≥99.0% (GC)
92741
4-Trimethylsiloxy-3-penten-2-one
13257-81-3
purum; ≥97.0% (GC)
92745
N-(Trimethylsilyl)acetamide
13435-12-6
pract.; ~90% (GC)
92753
Trimethylsilyl azide
4648-54-8
purum; ~97% (GC)
12700
Trimethylsilyl benzenesulfonate
17882-06-3
purum; ≥98.0% (T)
39860
Trimethylsilyl N,N-dimethylcarbamate
32115-55-2
purum; ~97% (GC)
92751
1-(Trimethylsilyl)imidazole
18156-74-6
purum; ≥98.0% (T)
92718
1-(Trimethylsilyl)imidazole/Pyridine1/4 (v/v), for GC
8077-35-8
for GC
64296
Trimethylsilyl methanesulfonate
10090-05-8
purum
77245
Trimethylsilyl nonafluoro-1-butane-sulfonate
68734-62-3
purum; ≥98.0% (T)
91741
Trimethylsilyl trifluoromethanesulfonate
27607-77-8
purum; ≥98.0% (T)
93133
Triphenylsilylamine
4215-80-9
purum; ~97% (NT)
Back to top
Acylating Reagents
Catalog No.
Product
CAS No.
Description
19667
Butylboronic acid, for GC
4426-47-5
BioChemika for GC; ~98% (T)
77251
Heptafluorobutyric anhydride, 10x1 ml
336-59-4
puriss.; ≥99.0% (GC)
77253
Heptafluorobutyric anhydride
336-59-4
puriss.; ≥99.0% (GC)
77254
Heptafluorobutyryl chloride
375-16-6
pract.; ≥98.0% (GC)
02379
2,3,4,5,6-Pentafluorobenzoic anhydride
15989-99-8
purum; ≥98.0% (GC)
77290
Pentafluoropropionic acid
422-64-0
purum; ≥97.0% (GC)
77292
Pentafluoropropionic anhydride
356-42-3
purum; ~98% (GC)
80903
Pivalic anhydride
1538-75-6
pract.; ≥97.0% (GC)
80910
Pivaloyl chloride
3282-30-2
purum; ≥98.0% (GC)
91719
Trifluoroacetic anhydride, derivatization reagent
407-25-0
BioChemika puriss. p.a. derivatization reagent for GC; ≥99.0% (GC)
91720
Trifluoroacetic anhydride
407-25-0
purum; ≥98.0% (GC)
Back to top
Alkylating and Esterifying Reagents
Catalog No.
Product
CAS No.
Description
15716
Boron trifluoride (~10% (~1.3 M) inMethanol)
373-57-9
puriss. for esterification of fatty acids for GC purposes
16460
4-Bromobenzyl bromide
589-15-1
purum; ≥98.0% (GC)
17910
alpha-Bromo-2,3,4,5,6-pentafluorotoluene
1765-40-8
puriss.; ≥99.0% (GC)
18790
alpha-Bromo-p-xylene
104-81-4
purum; ≥97.0% (GC)
40263
N,N-Dimethylformamide di-tert-butylacetal
36805-97-7
pract.; ≥90% (GC)
40252
N,N-Dimethylformamide diethyl acetal
1188-33-6
pract. for esterification of fatty acids; ~95% (GC)
40271
N,N-Dimethylformamide dimethyl acetal
4637-24-5
pract.; ≥95% (GC)
40680
N,N-Dimethylhydrazine (asym.) anhydrous
57-14-7
purum; ≥98.0% (GC)
52517
1,1,1,3,3,3-Hexafluoro-2-propanol
920-66-1
puriss. p.a. for GC; ≥99,8% (GC)
17934
Hydrogen chloride 1.25 M in Ethanol
123864-74-4
puriss. p.a. for GC; ~1.25 M (T)
17935
Hydrogen chloride 1.25 M in Methanol
132228-87-6
puriss. p.a. for GC; ~1.25 M (T)
17933
Hydrogen chloride 1.25 in 2-Propanol   puriss. p.a. for GC; ~1.25 M (T)
64954
2-Methoxy-1,3-dioxolane
19693-75-5
purum; ≥98.0% (GC)
66638
O-Methyl-N,N'-diisopropylisourea
54648-79-2
BioChemika purum; ≥97.0% (GC)
77256
Nonafluoro-1-iodobutane, stab.
423-39-2
purum; ≥98.0% (GC); stab. with silver wool
77265
Pentafluoro-iodoethane
354-64-3
purum; ≥97.0% (GC)
76757
2,2,3,3,3-Pentafluoro-1-propanol
422-05-9
purum; ≥97.0% (GC)
79265
Phenyltrimethylammonium hydroxide Sol.,~0.1 M in methanol
1899-02-1
puriss. p.a. for GC; ~0.1 M in methanol
79266
Phenyltrimethylammonium hydroxide Sol.,~0.5 M in methanol
1899-02-1
puriss. p.a. for GC; ~0.5 M in methanol
87285
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide
76437-40-6
purum; ≥97.0% (GC)
91120
2,2,2-Trichloroethanol
115-20-8
BioChemika pract.; ~95% (GC)
91690
2,2,2-Trifluoroethanol
75-89-8
puriss.; ≥99.0% (GC)
92334
Trimethylboroxine
823-96-1
puriss.; ≥99.0% (GC)
92732
Trimethylsulfonium hydrox. Sol., ~0.25 M in MeOH, 10x1 ml
17287-03-5
purum derivatization reagent for GC; ~0.25 M in methanol
Back to top
Back to GC Home